Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTNA
Entrez Gene ID
1837
Gene Name
dystrobrevin alpha
Gene Aliases
D18S892E, DRP3, DTN, DTN-A, LVNC1
Chromosome Location
Chr.18: 34493290 - 34891844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128175.1 NP_001121647.1 7 - 8 948 68
NM_001198938.1 NP_001185867.1 7 - 8 948 68
NM_001198939.1 NP_001185868.1 7 - 8 948 68
NM_001198940.1 NP_001185869.1 7 - 8 948 68
NM_001198941.1 NP_001185870.1 6 - 7 816 68
NM_001390.4 NP_001381.2 5 - 6 602 68
NM_001391.5 NP_001382.2 5 - 6 602 68
NM_001392.4 NP_001383.2 7 - 8 952 68
NM_032975.3 NP_116757.2 7 - 8 952 68
NM_032978.6 NP_116760.2 6 - 7 816 68
NM_032979.4 NP_116761.2 7 - 8 952 68
XM_005258221.4 XP_005258278.1 7 - 8 947 68
XM_011525853.2 XP_011524155.1 7 - 8 947 68
XM_017025575.1 XP_016881064.1 6 - 7 816 68
XM_017025576.1 XP_016881065.1 7 - 8 941 68
XM_017025577.1 XP_016881066.1 6 - 7 816 68
XM_017025578.1 XP_016881067.1 7 - 8 941 68
XM_017025579.1 XP_016881068.1 6 - 7 816 68
XM_017025580.1 XP_016881069.1 7 - 8 947 68
XM_017025581.1 XP_016881070.1 7 - 8 941 68
XM_017025582.1 XP_016881071.1 6 - 7 816 68
XM_017025583.1 XP_016881072.1 7 - 8 947 68
XM_017025584.1 XP_016881073.1 7 - 8 941 68
XM_017025585.1 XP_016881074.1 7 - 8 941 68
XM_017025586.1 XP_016881075.1 7 - 8 941 68
XM_017025587.1 XP_016881076.1 7 - 8 941 68
XM_017025588.1 XP_016881077.1 6 - 7 816 68
XM_017025589.1 XP_016881078.1 7 - 8 947 68
XM_017025590.1 XP_016881079.1 7 - 8 941 68
XM_017025591.1 XP_016881080.1 6 - 7 816 68
XM_017025592.1 XP_016881081.1 7 - 8 947 68
XM_017025593.1 XP_016881082.1 7 - 8 941 68
XM_017025594.1 XP_016881083.1 6 - 7 816 68
XM_017025595.1 XP_016881084.1 7 - 8 947 68
XM_017025596.1 XP_016881085.1 7 - 8 947 68
XM_017025597.1 XP_016881086.1 7 - 8 941 68
XM_017025598.1 XP_016881087.1 6 - 7 816 68
XM_017025600.1 XP_016881089.1 6 - 7 816 68
XM_017025601.1 XP_016881090.1 7 - 8 941 68
XM_017025602.1 XP_016881091.1 6 - 7 816 68
GenBank mRNA AB209102.1 - 7 - 8 901 68
AJ009668.1 - 7 - 8 860 68
AK291156.1 - 6 - 7 751 68
AK308410.1 - 7 - 8 928 68
AL833285.1 - 7 - 8 908 68
BC005300.1 - 7 - 8 908 68
BT006937.1 - 5 - 6 601 68
U26742.1 - 7 - 8 925 68
U26744.1 - 6 - 7 659 68
U46744.1 - 6 - 7 665 68
U46745.1 - 6 - 7 872 68

View Full Product Details