The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001621.4 | NP_001612.1 | 4 - 5 | 1061 | 118 |
GenBank mRNA | AK307495.1 | - | 4 - 5 | 1061 | 118 |
BC069390.1 | - | 4 - 5 | 518 | 118 | |
BC070080.1 | - | 4 - 5 | 979 | 118 | |
D16354.1 | - | 4 - 5 | 1030 | 118 | |
L19872.1 | - | 4 - 5 | 823 | 118 |