The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002111.8 | NP_002102.4 | 24 - 25 | 3298 | 81 |
GenBank mRNA | AB016794.1 | - | 24 - 25 | 3298 | 81 |
AB209506.1 | - | 11 - 12 | 3246 | 81 | |
L12392.1 | - | 24 - 25 | 3468 | 81 |