The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002111.8 | NP_002102.4 | 36 - 37 | 4900 | 73 |
GenBank mRNA | AB016794.1 | - | 36 - 37 | 4900 | 73 |
AB209506.1 | - | 23 - 24 | 4848 | 73 | |
L12392.1 | - | 36 - 37 | 5070 | 73 |