The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004260.3 | NP_004251.3 | 14 - 15 | 2510 | 69 |
XM_017013991.1 | XP_016869480.1 | 13 - 14 | 2605 | 69 | |
XM_017013992.1 | XP_016869481.1 | 13 - 14 | 2604 | 69 | |
XM_017013993.1 | XP_016869482.1 | 14 - 15 | 2514 | 69 | |
XM_017013994.1 | XP_016869483.1 | 14 - 15 | 2510 | 69 | |
XM_017013995.1 | XP_016869484.1 | 14 - 15 | 2512 | 69 | |
XM_017013996.1 | XP_016869485.1 | 13 - 14 | 2603 | 69 | |
XM_017013998.1 | XP_016869487.1 | 13 - 14 | 2602 | 69 | |
XM_017014000.1 | XP_016869489.1 | 9 - 10 | 1516 | 69 | |
XM_017014001.1 | XP_016869490.1 | 12 - 13 | 2606 | 69 | |
GenBank mRNA | AB006532.1 | - | 14 - 15 | 2468 | 69 |
BC011602.2 | - | 7 - 8 | 1069 | 69 | |
BC013277.2 | - | 5 - 6 | 807 | 69 |