Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39244725-39245231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNL3
Entrez Gene ID
282617
Gene Name
interferon lambda 3
Gene Aliases
IL-28B, IL28B, IL28C
UniGene
Hs.406744
Gene Chromosome Location
Chr.19: 39243536-39245481 on Build GRCh38
Transcripts
NM_172139.2 XM_005258765.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3223459 c.164G>A p.R55K Chr.19: 39244804-39244804 on Build GRCh38
1393513 c.155C>T p.A52V Chr.19: 39244813-39244813 on Build GRCh38
3756772 c.91C>T p.L31F Chr.19: 39244877-39244877 on Build GRCh38
1393512 c.161A>G p.K54R Chr.19: 39244807-39244807 on Build GRCh38
7423944 c.161A>T p.K54M Chr.19: 39244807-39244807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370170741 Chr.19: 39244916-39244916 on Build GRCh38
rs201412210 Chr.19: 39244850-39244850 on Build GRCh38
rs629976 Chr.19: 39244885-39244885 on Build GRCh38
rs1053882311 Chr.19: 39244777-39244777 on Build GRCh38
rs768807826 Chr.19: 39244838-39244838 on Build GRCh38
rs375329061 Chr.19: 39244835-39244835 on Build GRCh38
rs200835829 Chr.19: 39245003-39245003 on Build GRCh38
rs28416813 Chr.19: 39245004-39245004 on Build GRCh38
rs201680991 Chr.19: 39244889-39244889 on Build GRCh38
rs774510939 Chr.19: 39244841-39244841 on Build GRCh38
rs748387416 Chr.19: 39244940-39244940 on Build GRCh38
rs371708541 Chr.19: 39244796-39244796 on Build GRCh38
rs772555991 Chr.19: 39244904-39244905 on Build GRCh38
rs200508945 Chr.19: 39244863-39244863 on Build GRCh38
rs201540002 Chr.19: 39244884-39244884 on Build GRCh38
rs141536970 Chr.19: 39244895-39244895 on Build GRCh38
rs1009775316 Chr.19: 39244818-39244818 on Build GRCh38
rs778494193 Chr.19: 39244772-39244772 on Build GRCh38
rs754694535 Chr.19: 39244872-39244872 on Build GRCh38
rs921224516 Chr.19: 39245132-39245132 on Build GRCh38
rs199950179 Chr.19: 39244744-39244744 on Build GRCh38
rs200610163 Chr.19: 39245103-39245103 on Build GRCh38
rs372735927 Chr.19: 39244981-39244981 on Build GRCh38
rs373577185 Chr.19: 39244891-39244891 on Build GRCh38
rs201228830 Chr.19: 39244943-39244943 on Build GRCh38
rs202126177 Chr.19: 39244963-39244963 on Build GRCh38
rs143935261 Chr.19: 39244967-39244967 on Build GRCh38
rs201991079 Chr.19: 39244781-39244781 on Build GRCh38
rs199999531 Chr.19: 39244907-39244907 on Build GRCh38
rs748295128 Chr.19: 39244903-39244904 on Build GRCh38
rs578007730 Chr.19: 39245126-39245126 on Build GRCh38
rs71356849 Chr.19: 39245000-39245000 on Build GRCh38
rs562968930 Chr.19: 39244876-39244876 on Build GRCh38
rs549676428 Chr.19: 39244888-39244888 on Build GRCh38
rs141779226 Chr.19: 39244928-39244928 on Build GRCh38
rs201422642 Chr.19: 39245109-39245109 on Build GRCh38
rs750265243 Chr.19: 39244758-39244758 on Build GRCh38
rs1003985706 Chr.19: 39245038-39245038 on Build GRCh38
rs866777577 Chr.19: 39245008-39245008 on Build GRCh38
rs989508405 Chr.19: 39245192-39245192 on Build GRCh38
rs199878734 Chr.19: 39245136-39245136 on Build GRCh38
rs761823135 Chr.19: 39244745-39244745 on Build GRCh38
rs146179451 Chr.19: 39244762-39244762 on Build GRCh38
rs144430183 Chr.19: 39244908-39244908 on Build GRCh38
rs200139569 Chr.19: 39245064-39245064 on Build GRCh38
rs201278888 Chr.19: 39245175-39245175 on Build GRCh38
rs868769262 Chr.19: 39245169-39245169 on Build GRCh38
rs200133361 Chr.19: 39244917-39244917 on Build GRCh38
rs767306139 Chr.19: 39244747-39244747 on Build GRCh38
rs560602940 Chr.19: 39245184-39245184 on Build GRCh38
rs202028512 Chr.19: 39244901-39244901 on Build GRCh38
rs202067131 Chr.19: 39244768-39244768 on Build GRCh38
rs780497389 Chr.19: 39244924-39244924 on Build GRCh38
rs630388 Chr.19: 39244962-39244962 on Build GRCh38
rs763166362 Chr.19: 39244834-39244834 on Build GRCh38
rs753602639 Chr.19: 39244767-39244767 on Build GRCh38
rs201598821 Chr.19: 39245062-39245062 on Build GRCh38
rs201252721 Chr.19: 39244771-39244771 on Build GRCh38
rs200909158 Chr.19: 39244896-39244896 on Build GRCh38
rs202129388 Chr.19: 39245046-39245046 on Build GRCh38
rs200719718 Chr.19: 39245047-39245047 on Build GRCh38
rs775823318 Chr.19: 39244824-39244824 on Build GRCh38
rs199632578 Chr.19: 39245019-39245019 on Build GRCh38
rs141678286 Chr.19: 39244992-39244992 on Build GRCh38
rs199952257 Chr.19: 39244798-39244798 on Build GRCh38
rs747041025 Chr.19: 39244958-39244958 on Build GRCh38
rs200147727 Chr.19: 39244769-39244769 on Build GRCh38
rs377221199 Chr.19: 39244949-39244949 on Build GRCh38
rs150569967 Chr.19: 39244961-39244961 on Build GRCh38
rs201018323 Chr.19: 39244804-39244804 on Build GRCh38
rs138562018 Chr.19: 39244886-39244886 on Build GRCh38
rs775284197 Chr.19: 39244975-39244975 on Build GRCh38
rs766344202 Chr.19: 39244864-39244864 on Build GRCh38
rs770198147 Chr.19: 39244820-39244820 on Build GRCh38
rs761756652 Chr.19: 39244842-39244842 on Build GRCh38
rs140951294 Chr.19: 39244883-39244883 on Build GRCh38
rs369774544 Chr.19: 39244881-39244881 on Build GRCh38
rs202115118 Chr.19: 39244853-39244853 on Build GRCh38
rs71356848 Chr.19: 39244951-39244951 on Build GRCh38
rs377212766 Chr.19: 39244990-39244990 on Build GRCh38
rs751012686 Chr.19: 39244915-39244915 on Build GRCh38
rs200272719 Chr.19: 39244843-39244843 on Build GRCh38
rs201579699 Chr.19: 39244873-39244873 on Build GRCh38
rs778932906 Chr.19: 39244894-39244905 on Build GRCh38
rs200289435 Chr.19: 39244966-39244966 on Build GRCh38
rs753624869 Chr.19: 39244871-39244871 on Build GRCh38
rs772101792 Chr.19: 39244890-39244890 on Build GRCh38
rs138893424 Chr.19: 39244877-39244877 on Build GRCh38
rs771494831 Chr.19: 39244791-39244791 on Build GRCh38
rs148017150 Chr.19: 39244909-39244909 on Build GRCh38
rs913967857 Chr.19: 39245203-39245203 on Build GRCh38
rs201838861 Chr.19: 39244837-39244837 on Build GRCh38
rs756014514 Chr.19: 39244761-39244761 on Build GRCh38

View Full Product Details