Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM2
Entrez Gene ID
2912
Gene Name
glutamate metabotropic receptor 2
Gene Aliases
GLUR2, GPRC1B, MGLUR2, mGlu2
Chromosome Location
Chr.3: 51707033 - 51718616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000839.3 NP_000830.2 2 - 3 688 63
XM_011533635.1 XP_011531937.1 1 - 2 2359 63
XM_011533642.1 XP_011531944.1 1 - 2 2359 63
XM_017006271.1 XP_016861760.1 2 - 3 1166 63
GenBank mRNA AB209164.1 - 2 - 3 1178 63
AK289984.1 - 2 - 3 681 63
AK313056.1 - 2 - 3 665 63
AK315816.1 - 2 - 3 683 63
AY999299.1 - 1 - 2 485 63
BC113615.1 - 1 - 2 472 63
BC113619.1 - 1 - 2 472 63
BX648951.1 - 1 - 2 2128 63
EU432122.1 - 1 - 2 449 63
L35318.1 - 1 - 2 451 63

View Full Product Details