The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001447.2 | NP_001438.1 | 22 - 23 | 12529 | 112 |
XM_006714761.3 | XP_006714824.1 | 23 - 24 | 13100 | 112 | |
XM_011537600.2 | XP_011535902.1 | 25 - 26 | 12868 | 112 | |
XM_011537603.2 | XP_011535905.1 | 23 - 24 | 12801 | 112 | |
XM_017009224.1 | XP_016864713.1 | 25 - 26 | 12870 | 112 | |
XM_017009225.1 | XP_016864714.1 | 24 - 25 | 12712 | 112 | |
GenBank mRNA | AB160984.1 | - | 10 - 11 | 2839 | 112 |
AF231022.1 | - | 22 - 23 | 12529 | 112 | |
AL157443.1 | - | 1 - 2 | 95 | 112 | |
BC039143.2 | - | 5 - 6 | 112 |