The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003006.4 | NP_002997.2 | 1 - 2 | 164 | 73 |
GenBank mRNA | AK098315.1 | - | 1 - 2 | 164 | 73 |
AK127638.1 | - | 1 - 2 | 162 | 73 | |
AK225644.1 | - | 1 - 2 | 60 | 73 | |
AK290395.1 | - | 1 - 2 | 131 | 73 | |
AK295238.1 | - | 1 - 2 | 66 | 73 | |
AK298738.1 | - | 1 - 2 | 162 | 73 | |
AK300057.1 | - | 1 - 2 | 111 | 73 | |
BC029782.1 | - | 1 - 2 | 143 | 73 | |
U02297.1 | - | 1 - 2 | 63 | 73 |