The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_122031.1 | 1 - 1 | 901 | 63 | |
GenBank mRNA | AF050080.1 | - | 1 - 1 | 342 | 63 |
AF050081.1 | - | 1 - 1 | 924 | 63 | |
BC093751.1 | - | 1 - 1 | 367 | 63 | |
BC104849.1 | - | 1 - 1 | 367 | 63 |