Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54604261-54604767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_147161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3865803 c.1192C>T p.L398F Chr.1: 54604385-54604385 on Build GRCh38
3490860 c.1185G>A p.L395L Chr.1: 54604378-54604378 on Build GRCh38
3490861 c.1185G>A p.L395L Chr.1: 54604378-54604378 on Build GRCh38
5526165 c.1220C>T p.S407F Chr.1: 54604413-54604413 on Build GRCh38
5526166 c.1220C>T p.S407F Chr.1: 54604413-54604413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12038884 Chr.1: 54604357-54604357 on Build GRCh38
rs772571857 Chr.1: 54604310-54604310 on Build GRCh38
rs899017983 Chr.1: 54604564-54604564 on Build GRCh38
rs771459556 Chr.1: 54604412-54604412 on Build GRCh38
rs752807018 Chr.1: 54604386-54604386 on Build GRCh38
rs34148246 Chr.1: 54604416-54604416 on Build GRCh38
rs889793439 Chr.1: 54604487-54604487 on Build GRCh38
rs974617899 Chr.1: 54604676-54604676 on Build GRCh38
rs932680556 Chr.1: 54604491-54604491 on Build GRCh38
rs773648115 Chr.1: 54604311-54604311 on Build GRCh38
rs374287351 Chr.1: 54604478-54604478 on Build GRCh38
rs548810628 Chr.1: 54604643-54604643 on Build GRCh38
rs980265631 Chr.1: 54604608-54604608 on Build GRCh38
rs1051569598 Chr.1: 54604481-54604481 on Build GRCh38
rs950664898 Chr.1: 54604583-54604583 on Build GRCh38
rs760646481 Chr.1: 54604456-54604456 on Build GRCh38
rs892302856 Chr.1: 54604552-54604552 on Build GRCh38
rs761357493 Chr.1: 54604443-54604443 on Build GRCh38
rs753629000 Chr.1: 54604477-54604477 on Build GRCh38
rs750365673 Chr.1: 54604351-54604351 on Build GRCh38
rs750287752 Chr.1: 54604507-54604507 on Build GRCh38
rs763663357 Chr.1: 54604327-54604327 on Build GRCh38
rs755948750 Chr.1: 54604362-54604362 on Build GRCh38
rs982986805 Chr.1: 54604406-54604406 on Build GRCh38
rs781469724 Chr.1: 54604415-54604415 on Build GRCh38
rs868391071 Chr.1: 54604607-54604607 on Build GRCh38
rs376188034 Chr.1: 54604398-54604398 on Build GRCh38
rs994607396 Chr.1: 54604566-54604566 on Build GRCh38
rs767487500 Chr.1: 54604350-54604350 on Build GRCh38
rs1052270367 Chr.1: 54604548-54604548 on Build GRCh38
rs79180715 Chr.1: 54604614-54604614 on Build GRCh38
rs564222412 Chr.1: 54604518-54604518 on Build GRCh38
rs983037662 Chr.1: 54604635-54604635 on Build GRCh38
rs753713724 Chr.1: 54604371-54604371 on Build GRCh38
rs950405727 Chr.1: 54604323-54604323 on Build GRCh38
rs746503 Chr.1: 54604313-54604313 on Build GRCh38
rs773944535 Chr.1: 54604439-54604439 on Build GRCh38
rs376969020 Chr.1: 54604451-54604451 on Build GRCh38
rs747467378 Chr.1: 54604409-54604409 on Build GRCh38
rs369221942 Chr.1: 54604329-54604329 on Build GRCh38
rs980818443 Chr.1: 54604352-54604352 on Build GRCh38
rs762553144 Chr.1: 54604322-54604322 on Build GRCh38
rs543001268 Chr.1: 54604603-54604603 on Build GRCh38
rs772867824 Chr.1: 54604454-54604454 on Build GRCh38
rs987034274 Chr.1: 54604642-54604642 on Build GRCh38
rs766223820 Chr.1: 54604367-54604367 on Build GRCh38
rs779016755 Chr.1: 54604383-54604383 on Build GRCh38
rs912571085 Chr.1: 54604730-54604730 on Build GRCh38
rs768346039 Chr.1: 54604417-54604417 on Build GRCh38
rs755264380 Chr.1: 54604713-54604713 on Build GRCh38
rs759304517 Chr.1: 54604364-54604365 on Build GRCh38
rs146094962 Chr.1: 54604425-54604425 on Build GRCh38
rs748181110 Chr.1: 54604306-54604306 on Build GRCh38
rs1034679817 Chr.1: 54604627-54604627 on Build GRCh38
rs963144706 Chr.1: 54604312-54604312 on Build GRCh38
rs761290500 Chr.1: 54604348-54604348 on Build GRCh38
rs771334853 Chr.1: 54604314-54604314 on Build GRCh38
rs376656975 Chr.1: 54604296-54604296 on Build GRCh38
rs533087364 Chr.1: 54604580-54604580 on Build GRCh38
rs200872982 Chr.1: 54604380-54604380 on Build GRCh38
rs180725204 Chr.1: 54604720-54604720 on Build GRCh38
rs913387788 Chr.1: 54604638-54604638 on Build GRCh38
rs369552793 Chr.1: 54604465-54604465 on Build GRCh38
rs750896281 Chr.1: 54604581-54604581 on Build GRCh38
rs930049387 Chr.1: 54604702-54604702 on Build GRCh38
rs373209712 Chr.1: 54604384-54604384 on Build GRCh38
rs528673743 Chr.1: 54604699-54604699 on Build GRCh38
rs752534671 Chr.1: 54604375-54604377 on Build GRCh38
rs867213752 Chr.1: 54604695-54604695 on Build GRCh38
rs928232750 Chr.1: 54604366-54604366 on Build GRCh38
rs183977443 Chr.1: 54604630-54604630 on Build GRCh38
rs371674143 Chr.1: 54604317-54604317 on Build GRCh38
rs1038423274 Chr.1: 54604557-54604557 on Build GRCh38
rs1051081995 Chr.1: 54604727-54604727 on Build GRCh38
rs1014164893 Chr.1: 54604590-54604590 on Build GRCh38
rs890343364 Chr.1: 54604288-54604288 on Build GRCh38
rs148824909 Chr.1: 54604495-54604495 on Build GRCh38
rs189252120 Chr.1: 54604658-54604658 on Build GRCh38
rs766379351 Chr.1: 54604656-54604656 on Build GRCh38
rs767509985 Chr.1: 54604369-54604371 on Build GRCh38
rs758472751 Chr.1: 54604395-54604395 on Build GRCh38
rs890412242 Chr.1: 54604505-54604505 on Build GRCh38

View Full Product Details