The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022468.4 | NP_071913.1 | 3 - 4 | 605 | 110 |
XM_011522602.2 | XP_011520904.1 | 3 - 4 | 342 | 110 | |
XM_011522604.2 | XP_011520906.1 | 2 - 3 | 456 | 110 | |
XM_017023561.1 | XP_016879050.1 | 3 - 4 | 1323 | 110 | |
GenBank mRNA | AB042328.1 | - | 3 - 4 | 537 | 110 |
AF145442.2 | - | 3 - 4 | 392 | 110 | |
AF185270.1 | - | 3 - 4 | 392 | 110 | |
AJ003144.1 | - | 2 - 3 | 153 | 110 | |
AJ239053.1 | - | 3 - 4 | 549 | 110 | |
AJ272137.1 | - | 3 - 4 | 605 | 110 |