The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004398.3 | NP_004389.2 | 8 - 9 | 1266 | 102 |
GenBank mRNA | AB040537.1 | - | 8 - 9 | 1202 | 102 |
AB209884.1 | - | 8 - 9 | 1169 | 102 | |
AK296752.1 | - | 6 - 7 | 940 | 102 | |
AK315216.1 | - | 8 - 9 | 1197 | 102 | |
BC049217.1 | - | 8 - 9 | 1162 | 102 | |
BC091521.1 | - | 8 - 9 | 1214 | 102 | |
BC093654.1 | - | 8 - 9 | 1170 | 102 | |
BC093656.1 | - | 8 - 9 | 1170 | 102 | |
U28042.1 | - | 8 - 9 | 1202 | 102 |