The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005641.3 | NP_005632.1 | 1 - 2 | 492 | 110 |
XM_017012568.1 | XP_016868057.1 | 1 - 2 | 492 | 110 | |
GenBank mRNA | AK314428.1 | - | 1 - 2 | 469 | 110 |
DA707589.1 | - | 1 - 2 | 494 | 110 | |
L25444.1 | - | 1 - 2 | 464 | 110 |