The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190415.1 | NP_001177344.1 | 11 - 12 | 1378 | 119 |
NM_005641.3 | NP_005632.1 | 11 - 12 | 1707 | 119 | |
NM_139315.2 | NP_647476.1 | 11 - 12 | 1409 | 119 | |
NR_033792.1 | 11 - 12 | 1258 | 119 | ||
XM_006716100.2 | XP_006716163.1 | 12 - 13 | 1500 | 119 | |
XM_006716101.2 | XP_006716164.1 | 11 - 12 | 1337 | 119 | |
XM_006716102.2 | XP_006716165.1 | 11 - 12 | 1204 | 119 | |
XM_011516542.2 | XP_011514844.1 | 12 - 13 | 1479 | 119 | |
XM_011516543.2 | XP_011514845.1 | 11 - 12 | 1232 | 119 | |
XM_017012568.1 | XP_016868057.1 | 11 - 12 | 1677 | 119 | |
GenBank mRNA | AK097781.1 | - | 11 - 12 | 1260 | 119 |
AK300005.1 | - | 11 - 12 | 1361 | 119 | |
AK314428.1 | - | 11 - 12 | 1684 | 119 | |
AK315077.1 | - | 11 - 12 | 1359 | 119 | |
BC018115.1 | - | 11 - 12 | 1352 | 119 | |
CR627390.1 | - | 11 - 12 | 1236 | 119 | |
L25444.1 | - | 11 - 12 | 1679 | 119 | |
U31659.1 | - | 10 - 11 | 1463 | 119 |