Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111145833-111146331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L11
Entrez Gene ID
10018
Gene Name
BCL2 like 11
Gene Aliases
BAM, BIM, BOD
UniGene
Hs.469658
Gene Chromosome Location
Chr.2: 111120914-111168445 on Build GRCh38
Transcripts
NM_001204106.1 NM_001204107.1 NM_001204108.1 NM_001204109.1 NM_001204110.1 NM_001204111.1 NM_001204112.1 NM_001204113.1 NM_006538.4 NM_138621.4 NM_138622.3 NM_138623.3 NM_138624.3 NM_138625.3 NM_138626.3 NM_138627.3 NM_207003.2 XM_005263550.3 XM_005263551.2 XM_005263552.3 XM_005263553.2 XM_005263554.2 XM_005263555.3 XM_005263556.3 XM_005263557.4 XM_005263559.2 XM_005263561.3 XM_017003101.1 XM_017003102.1

SNP Detail

SNP ID Chromosome Location
rs1050264574 Chr.2: 111146071-111146075 on Build GRCh38
rs892520126 Chr.2: 111146087-111146087 on Build GRCh38
rs904471603 Chr.2: 111146058-111146058 on Build GRCh38
rs76141679 Chr.2: 111146291-111146291 on Build GRCh38
rs1050040616 Chr.2: 111146299-111146299 on Build GRCh38
rs1009619430 Chr.2: 111146096-111146096 on Build GRCh38
rs770316710 Chr.2: 111146023-111146023 on Build GRCh38
rs937473678 Chr.2: 111145865-111145866 on Build GRCh38
rs984678564 Chr.2: 111146176-111146176 on Build GRCh38
rs898803460 Chr.2: 111146003-111146003 on Build GRCh38
rs866528269 Chr.2: 111145925-111145925 on Build GRCh38
rs186322907 Chr.2: 111146302-111146302 on Build GRCh38
rs1009270134 Chr.2: 111146103-111146103 on Build GRCh38
rs931469869 Chr.2: 111145924-111145924 on Build GRCh38
rs551024779 Chr.2: 111146112-111146112 on Build GRCh38
rs561794446 Chr.2: 111145882-111145882 on Build GRCh38
rs1034760087 Chr.2: 111145920-111145920 on Build GRCh38
rs545079974 Chr.2: 111146087-111146087 on Build GRCh38
rs953021082 Chr.2: 111146172-111146172 on Build GRCh38
rs1056796673 Chr.2: 111145961-111145961 on Build GRCh38
rs965571281 Chr.2: 111145888-111145888 on Build GRCh38
rs908275952 Chr.2: 111145926-111145926 on Build GRCh38
rs79951792 Chr.2: 111145941-111145941 on Build GRCh38
rs975643034 Chr.2: 111145907-111145907 on Build GRCh38
rs935950026 Chr.2: 111146249-111146249 on Build GRCh38
rs78947792 Chr.2: 111146254-111146254 on Build GRCh38
rs1028349819 Chr.2: 111146305-111146305 on Build GRCh38
rs1000421739 Chr.2: 111146061-111146061 on Build GRCh38
rs939738617 Chr.2: 111146192-111146192 on Build GRCh38
rs180684741 Chr.2: 111146167-111146167 on Build GRCh38
rs965351405 Chr.2: 111146161-111146161 on Build GRCh38
rs73954958 Chr.2: 111146165-111146165 on Build GRCh38
rs1044111096 Chr.2: 111146243-111146243 on Build GRCh38
rs1019589994 Chr.2: 111145886-111145886 on Build GRCh38
rs114039165 Chr.2: 111146306-111146306 on Build GRCh38
rs921393182 Chr.2: 111145919-111145919 on Build GRCh38
rs992587059 Chr.2: 111146200-111146200 on Build GRCh38
rs904418035 Chr.2: 111146248-111146248 on Build GRCh38
rs150272389 Chr.2: 111146272-111146272 on Build GRCh38
rs202067352 Chr.2: 111145923-111145924 on Build GRCh38
rs917410501 Chr.2: 111146224-111146224 on Build GRCh38
rs927533874 Chr.2: 111145863-111145863 on Build GRCh38
rs3761705 Chr.2: 111145950-111145950 on Build GRCh38
rs3761704 Chr.2: 111146121-111146121 on Build GRCh38
rs58738963 Chr.2: 111145936-111145940 on Build GRCh38
rs895498599 Chr.2: 111145968-111145968 on Build GRCh38
rs548267886 Chr.2: 111145881-111145881 on Build GRCh38
rs1010294373 Chr.2: 111146158-111146158 on Build GRCh38
rs1031525811 Chr.2: 111146066-111146066 on Build GRCh38
rs749626028 Chr.2: 111146222-111146222 on Build GRCh38
rs760188940 Chr.2: 111146111-111146111 on Build GRCh38
rs74648438 Chr.2: 111146303-111146303 on Build GRCh38
rs528572698 Chr.2: 111145865-111145865 on Build GRCh38
rs138780074 Chr.2: 111145920-111145920 on Build GRCh38
rs371946113 Chr.2: 111145992-111145992 on Build GRCh38
rs908729618 Chr.2: 111146184-111146184 on Build GRCh38
rs1057291592 Chr.2: 111145911-111145911 on Build GRCh38
rs1044339547 Chr.2: 111146036-111146036 on Build GRCh38
rs956778349 Chr.2: 111146146-111146146 on Build GRCh38
rs567032126 Chr.2: 111146236-111146236 on Build GRCh38
rs11674517 Chr.2: 111146255-111146255 on Build GRCh38
rs1009566991 Chr.2: 111146266-111146266 on Build GRCh38

View Full Product Details