Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113062737-113063234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL36RN
Entrez Gene ID
26525
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
FIL1, FIL1(DELTA), FIL1D, IL-36Ra, IL1F5, IL1HY1, IL1L1, IL1RP3, IL36RA, PSORP, PSORS14
UniGene
Hs.516301
Gene Chromosome Location
Chr.2: 113058638-113064744 on Build GRCh38
Transcripts
NM_012275.2 NM_173170.1

SNP Detail

SNP ID Chromosome Location
rs3207835 Chr.2: 113062782-113062782 on Build GRCh38
rs1023058124 Chr.2: 113063199-113063199 on Build GRCh38
rs573648177 Chr.2: 113063030-113063030 on Build GRCh38
rs182598494 Chr.2: 113062783-113062783 on Build GRCh38
rs2515402 Chr.2: 113063003-113063003 on Build GRCh38
rs3171809 Chr.2: 113063082-113063082 on Build GRCh38
rs893347733 Chr.2: 113063176-113063176 on Build GRCh38
rs565298108 Chr.2: 113063190-113063190 on Build GRCh38
rs113980876 Chr.2: 113062996-113062996 on Build GRCh38
rs984138108 Chr.2: 113062822-113062822 on Build GRCh38
rs114200724 Chr.2: 113062985-113062985 on Build GRCh38
rs925819174 Chr.2: 113062974-113062974 on Build GRCh38
rs3207836 Chr.2: 113062832-113062832 on Build GRCh38
rs1011800928 Chr.2: 113063185-113063185 on Build GRCh38
rs780966265 Chr.2: 113062984-113062984 on Build GRCh38
rs3180234 Chr.2: 113063078-113063078 on Build GRCh38
rs915665674 Chr.2: 113062951-113062951 on Build GRCh38
rs1022642407 Chr.2: 113063204-113063204 on Build GRCh38
rs187067535 Chr.2: 113063021-113063021 on Build GRCh38
rs529172925 Chr.2: 113062842-113062842 on Build GRCh38
rs2515403 Chr.2: 113063155-113063155 on Build GRCh38
rs3207839 Chr.2: 113062971-113062971 on Build GRCh38
rs3207837 Chr.2: 113062862-113062862 on Build GRCh38
rs78047571 Chr.2: 113063090-113063090 on Build GRCh38
rs1800930 Chr.2: 113062953-113062953 on Build GRCh38
rs934678828 Chr.2: 113063107-113063107 on Build GRCh38
rs980557987 Chr.2: 113062926-113062926 on Build GRCh38
rs900605689 Chr.2: 113063012-113063012 on Build GRCh38
rs1039098006 Chr.2: 113063010-113063010 on Build GRCh38
rs149933228 Chr.2: 113063086-113063086 on Build GRCh38
rs1023089122 Chr.2: 113062876-113062876 on Build GRCh38
rs547397492 Chr.2: 113062776-113062776 on Build GRCh38
rs80220660 Chr.2: 113062958-113062958 on Build GRCh38
rs3180235 Chr.2: 113063095-113063095 on Build GRCh38
rs2515401 Chr.2: 113062899-113062899 on Build GRCh38
rs748951274 Chr.2: 113063096-113063096 on Build GRCh38
rs139175000 Chr.2: 113062935-113062935 on Build GRCh38
rs994974881 Chr.2: 113063019-113063019 on Build GRCh38

View Full Product Details