The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016270.3 | NP_057354.1 | 1 - 2 | 177 | 116 |
GenBank mRNA | AF205849.1 | - | 1 - 2 | 163 | 116 |
AK225820.1 | - | 1 - 2 | 177 | 116 | |
BC035342.1 | - | 1 - 2 | 164 | 116 | |
BC071983.1 | - | 1 - 2 | 166 | 116 | |
CA950627.1 | - | 1 - 2 | 115 | 116 |