Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BEX1
Entrez Gene ID
55859
Gene Name
brain expressed X-linked 1
Gene Aliases
BEX2, HBEX2, HGR74-h
Chromosome Location
Chr.X: 103062653 - 103064240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018476.3 NP_060946.3 1 - 2 141 111
GenBank mRNA AF237783.1 - 1 - 2 72 111
AK290958.1 - 1 - 2 75 111
BC105298.1 - 1 - 2 76 111
BM465948.1 - 1 - 2 82 111
BX098763.1 - 1 - 2 141 111

View Full Product Details