Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APOBR
Entrez Gene ID
55911
Gene Name
apolipoprotein B receptor
Gene Aliases
APOB100R, APOB48R
Chromosome Location
Chr.16: 28494649 - 28498970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018690.3 NP_061160.3 3 - 4 3257 84
GenBank mRNA AF141332.1 - 4 - 5 3207 84
AF141334.1 - 4 - 5 3207 84
AK025123.1 - 2 - 3 1109 84
BC119786.1 - 4 - 5 3207 84
BC119788.1 - 4 - 5 3002 84

View Full Product Details