The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317895.1 | NP_001304824.1 | 6 - 7 | 2014 | 74 |
NM_032789.4 | NP_116178.2 | 7 - 8 | 1890 | 74 | |
NR_134234.1 | 7 - 8 | 1890 | 74 | ||
GenBank mRNA | AK027370.1 | - | 7 - 8 | 1847 | 74 |
AK075250.1 | - | 3 - 4 | 577 | 74 | |
AK128537.1 | - | 8 - 9 | 1705 | 74 | |
AK222914.1 | - | 7 - 8 | 1873 | 74 | |
AK303317.1 | - | 6 - 7 | 1939 | 74 | |
BC019030.2 | - | 3 - 4 | 710 | 74 | |
BC030549.1 | - | 7 - 8 | 1864 | 74 | |
BC132961.1 | - | 7 - 8 | 1841 | 74 | |
BC136542.1 | - | 7 - 8 | 1841 | 74 | |
BC144235.1 | - | 7 - 8 | 1841 | 74 |