Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17120419-17120905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR6
Entrez Gene ID
27439
Gene Name
cat eye syndrome chromosome region, candidate 6
Gene Aliases
-
UniGene
Hs.209577
Gene Chromosome Location
Chr.22: 17116299-17121367 on Build GRCh38
Transcripts
NM_001163079.1 NM_031890.3 XM_011546124.2 XM_011546125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7424570 c.638G>T p.G213V Chr.22: 17120490-17120490 on Build GRCh38
6036966 c.619C>G p.L207V Chr.22: 17120509-17120509 on Build GRCh38
3800011 c.690G>A p.W230* Chr.22: 17120438-17120438 on Build GRCh38
7360740 c.670G>A p.D224N Chr.22: 17120458-17120458 on Build GRCh38
5834076 c.625_627delCTG p.L209delL Chr.22: 17120501-17120503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141276331 Chr.22: 17120475-17120475 on Build GRCh38
rs1057389475 Chr.22: 17120641-17120641 on Build GRCh38
rs766319866 Chr.22: 17120485-17120485 on Build GRCh38
rs762336989 Chr.22: 17120598-17120598 on Build GRCh38
rs897431469 Chr.22: 17120661-17120661 on Build GRCh38
rs756934137 Chr.22: 17120549-17120549 on Build GRCh38
rs915085990 Chr.22: 17120787-17120787 on Build GRCh38
rs897412215 Chr.22: 17120850-17120850 on Build GRCh38
rs1048508045 Chr.22: 17120885-17120885 on Build GRCh38
rs746245224 Chr.22: 17120553-17120553 on Build GRCh38
rs776603085 Chr.22: 17120486-17120486 on Build GRCh38
rs752141985 Chr.22: 17120504-17120504 on Build GRCh38
rs759468510 Chr.22: 17120581-17120581 on Build GRCh38
rs561363364 Chr.22: 17120580-17120580 on Build GRCh38
rs774410086 Chr.22: 17120573-17120573 on Build GRCh38
rs774694938 Chr.22: 17120471-17120471 on Build GRCh38
rs772432773 Chr.22: 17120578-17120578 on Build GRCh38
rs768832803 Chr.22: 17120566-17120566 on Build GRCh38
rs1053648548 Chr.22: 17120586-17120586 on Build GRCh38
rs1006601271 Chr.22: 17120696-17120696 on Build GRCh38
rs915451138 Chr.22: 17120818-17120818 on Build GRCh38
rs757982369 Chr.22: 17120529-17120529 on Build GRCh38
rs765267904 Chr.22: 17120498-17120498 on Build GRCh38
rs989655902 Chr.22: 17120684-17120684 on Build GRCh38
rs767161496 Chr.22: 17120773-17120773 on Build GRCh38
rs1057448017 Chr.22: 17120849-17120849 on Build GRCh38
rs768757723 Chr.22: 17120465-17120465 on Build GRCh38
rs1003514001 Chr.22: 17120640-17120640 on Build GRCh38
rs1027151525 Chr.22: 17120681-17120681 on Build GRCh38
rs914641222 Chr.22: 17120587-17120587 on Build GRCh38
rs201443358 Chr.22: 17120548-17120548 on Build GRCh38
rs756548327 Chr.22: 17120557-17120557 on Build GRCh38
rs952454225 Chr.22: 17120695-17120695 on Build GRCh38
rs952490239 Chr.22: 17120547-17120547 on Build GRCh38
rs780680596 Chr.22: 17120558-17120558 on Build GRCh38
rs965090269 Chr.22: 17120711-17120711 on Build GRCh38
rs377534248 Chr.22: 17120454-17120454 on Build GRCh38
rs760368856 Chr.22: 17120479-17120479 on Build GRCh38
rs947487700 Chr.22: 17120589-17120589 on Build GRCh38
rs994279888 Chr.22: 17120665-17120665 on Build GRCh38
rs938685169 Chr.22: 17120845-17120845 on Build GRCh38
rs759485113 Chr.22: 17120492-17120492 on Build GRCh38
rs165805 Chr.22: 17120744-17120744 on Build GRCh38
rs749317723 Chr.22: 17120560-17120560 on Build GRCh38
rs947413924 Chr.22: 17120804-17120804 on Build GRCh38
rs1044757396 Chr.22: 17120601-17120601 on Build GRCh38
rs749391815 Chr.22: 17120464-17120464 on Build GRCh38
rs769838837 Chr.22: 17120582-17120582 on Build GRCh38
rs927259015 Chr.22: 17120842-17120842 on Build GRCh38
rs75243280 Chr.22: 17120576-17120576 on Build GRCh38
rs923866621 Chr.22: 17120734-17120734 on Build GRCh38
rs763630817 Chr.22: 17120539-17120539 on Build GRCh38
rs763619025 Chr.22: 17120611-17120611 on Build GRCh38
rs751053503 Chr.22: 17120627-17120627 on Build GRCh38
rs1044504768 Chr.22: 17120817-17120817 on Build GRCh38
rs989310098 Chr.22: 17120781-17120783 on Build GRCh38
rs775545515 Chr.22: 17120585-17120585 on Build GRCh38
rs9606620 Chr.22: 17120858-17120858 on Build GRCh38
rs1018317041 Chr.22: 17120706-17120706 on Build GRCh38
rs573336073 Chr.22: 17120750-17120750 on Build GRCh38
rs781488099 Chr.22: 17120552-17120552 on Build GRCh38
rs977045235 Chr.22: 17120725-17120725 on Build GRCh38

View Full Product Details