The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003386.2 | NP_003377.2 | 36 - 37 | 7000 | 76 |
NM_173059.2 | NP_775082.2 | 36 - 37 | 7000 | 76 | |
NR_111917.1 | 36 - 37 | 7000 | 76 | ||
NR_111918.1 | 36 - 37 | 7000 | 76 | ||
NR_111919.1 | 36 - 37 | 7000 | 76 | ||
XM_011516555.2 | XP_011514857.1 | 36 - 37 | 6733 | 76 | |
XM_017012581.1 | XP_016868070.1 | 23 - 24 | 4489 | 76 | |
GenBank mRNA | AF332975.1 | - | 36 - 37 | 7000 | 76 |
AF332976.1 | - | 36 - 37 | 7000 | 76 | |
AF332977.1 | - | 36 - 37 | 7000 | 76 | |
AF332978.1 | - | 36 - 37 | 7000 | 76 | |
AF332979.1 | - | 36 - 37 | 7000 | 76 | |
AF332980.1 | - | 36 - 37 | 7000 | 76 | |
AK302596.1 | - | 15 - 16 | 2319 | 76 |