Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20976129-20976619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6296599 c.818G>T p.G273V Chr.22: 20976261-20976261 on Build GRCh38
5456934 c.956C>T p.T319M Chr.22: 20976464-20976464 on Build GRCh38
5495242 c.980G>T p.G327V Chr.22: 20976524-20976524 on Build GRCh38
5495240 c.1016G>T p.G339V Chr.22: 20976524-20976524 on Build GRCh38
5456935 c.974C>T p.T325M Chr.22: 20976464-20976464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377284602 Chr.22: 20976457-20976457 on Build GRCh38
rs533454844 Chr.22: 20976161-20976161 on Build GRCh38
rs369724765 Chr.22: 20976531-20976531 on Build GRCh38
rs764505179 Chr.22: 20976459-20976459 on Build GRCh38
rs930475653 Chr.22: 20976419-20976419 on Build GRCh38
rs753411995 Chr.22: 20976516-20976516 on Build GRCh38
rs570348375 Chr.22: 20976536-20976536 on Build GRCh38
rs1044389489 Chr.22: 20976345-20976345 on Build GRCh38
rs371705249 Chr.22: 20976230-20976230 on Build GRCh38
rs376144193 Chr.22: 20976162-20976181 on Build GRCh38
rs773595892 Chr.22: 20976399-20976399 on Build GRCh38
rs780154816 Chr.22: 20976221-20976221 on Build GRCh38
rs775711428 Chr.22: 20976575-20976575 on Build GRCh38
rs759716867 Chr.22: 20976503-20976503 on Build GRCh38
rs747656974 Chr.22: 20976483-20976483 on Build GRCh38
rs897657891 Chr.22: 20976149-20976149 on Build GRCh38
rs150801089 Chr.22: 20976461-20976461 on Build GRCh38
rs751979551 Chr.22: 20976182-20976182 on Build GRCh38
rs766601469 Chr.22: 20976447-20976447 on Build GRCh38
rs533580715 Chr.22: 20976520-20976520 on Build GRCh38
rs774599447 Chr.22: 20976337-20976337 on Build GRCh38
rs760681139 Chr.22: 20976522-20976522 on Build GRCh38
rs762030241 Chr.22: 20976194-20976194 on Build GRCh38
rs763269684 Chr.22: 20976169-20976179 on Build GRCh38
rs1047490293 Chr.22: 20976189-20976189 on Build GRCh38
rs7285694 Chr.22: 20976498-20976498 on Build GRCh38
rs776021522 Chr.22: 20976167-20976183 on Build GRCh38
rs376361841 Chr.22: 20976232-20976232 on Build GRCh38
rs901622675 Chr.22: 20976552-20976552 on Build GRCh38
rs769223224 Chr.22: 20976310-20976310 on Build GRCh38
rs376606947 Chr.22: 20976559-20976559 on Build GRCh38
rs754639841 Chr.22: 20976479-20976479 on Build GRCh38
rs900361173 Chr.22: 20976596-20976596 on Build GRCh38
rs751903383 Chr.22: 20976460-20976460 on Build GRCh38
rs1019260906 Chr.22: 20976423-20976423 on Build GRCh38
rs369535222 Chr.22: 20976510-20976510 on Build GRCh38
rs376395658 Chr.22: 20976321-20976321 on Build GRCh38
rs190234792 Chr.22: 20976566-20976566 on Build GRCh38
rs776650994 Chr.22: 20976448-20976448 on Build GRCh38
rs181636008 Chr.22: 20976500-20976500 on Build GRCh38
rs748955787 Chr.22: 20976476-20976476 on Build GRCh38
rs769487929 Chr.22: 20976165-20976165 on Build GRCh38
rs199698549 Chr.22: 20976218-20976218 on Build GRCh38
rs748649192 Chr.22: 20976395-20976395 on Build GRCh38
rs781123102 Chr.22: 20976257-20976257 on Build GRCh38
rs752164322 Chr.22: 20976534-20976534 on Build GRCh38
rs968172682 Chr.22: 20976313-20976313 on Build GRCh38
rs778330092 Chr.22: 20976482-20976482 on Build GRCh38
rs200032972 Chr.22: 20976517-20976517 on Build GRCh38
rs376210698 Chr.22: 20976371-20976371 on Build GRCh38
rs769919532 Chr.22: 20976569-20976569 on Build GRCh38
rs185338332 Chr.22: 20976511-20976511 on Build GRCh38
rs751509553 Chr.22: 20976350-20976350 on Build GRCh38
rs763259745 Chr.22: 20976576-20976576 on Build GRCh38
rs764475627 Chr.22: 20976317-20976319 on Build GRCh38
rs757196997 Chr.22: 20976359-20976359 on Build GRCh38
rs557227671 Chr.22: 20976471-20976471 on Build GRCh38
rs747563522 Chr.22: 20976549-20976549 on Build GRCh38
rs968679291 Chr.22: 20976481-20976481 on Build GRCh38
rs201100852 Chr.22: 20976248-20976248 on Build GRCh38
rs202225684 Chr.22: 20976544-20976544 on Build GRCh38
rs1027393802 Chr.22: 20976160-20976160 on Build GRCh38
rs750706750 Chr.22: 20976201-20976201 on Build GRCh38
rs745566482 Chr.22: 20976268-20976268 on Build GRCh38
rs370587105 Chr.22: 20976383-20976383 on Build GRCh38
rs930365053 Chr.22: 20976178-20976178 on Build GRCh38
rs374046191 Chr.22: 20976394-20976394 on Build GRCh38
rs769542944 Chr.22: 20976293-20976293 on Build GRCh38
rs760980659 Chr.22: 20976430-20976430 on Build GRCh38
rs201655926 Chr.22: 20976499-20976499 on Build GRCh38
rs748700543 Chr.22: 20976306-20976306 on Build GRCh38
rs569509463 Chr.22: 20976229-20976229 on Build GRCh38
rs141390776 Chr.22: 20976247-20976247 on Build GRCh38
rs749409732 Chr.22: 20976349-20976349 on Build GRCh38
rs149430218 Chr.22: 20976489-20976489 on Build GRCh38
rs762035787 Chr.22: 20976416-20976416 on Build GRCh38
rs34766252 Chr.22: 20976319-20976319 on Build GRCh38
rs369989227 Chr.22: 20976332-20976332 on Build GRCh38
rs772592484 Chr.22: 20976484-20976484 on Build GRCh38
rs778581948 Chr.22: 20976533-20976533 on Build GRCh38
rs140466618 Chr.22: 20976529-20976529 on Build GRCh38
rs755902017 Chr.22: 20976465-20976465 on Build GRCh38
rs888968075 Chr.22: 20976435-20976435 on Build GRCh38
rs147200843 Chr.22: 20976506-20976506 on Build GRCh38
rs756313045 Chr.22: 20976212-20976212 on Build GRCh38
rs766141089 Chr.22: 20976528-20976528 on Build GRCh38
rs139938405 Chr.22: 20976341-20976341 on Build GRCh38
rs752960303 Chr.22: 20976276-20976276 on Build GRCh38
rs372650074 Chr.22: 20976515-20976515 on Build GRCh38
rs764471755 Chr.22: 20976181-20976181 on Build GRCh38
rs781174868 Chr.22: 20976360-20976360 on Build GRCh38
rs367609112 Chr.22: 20976164-20976164 on Build GRCh38
rs995113867 Chr.22: 20976150-20976150 on Build GRCh38
rs139810844 Chr.22: 20976464-20976464 on Build GRCh38
rs773540672 Chr.22: 20976327-20976327 on Build GRCh38
rs369333874 Chr.22: 20976180-20976180 on Build GRCh38
rs961355962 Chr.22: 20976505-20976505 on Build GRCh38
rs765355336 Chr.22: 20976348-20976348 on Build GRCh38
rs745621478 Chr.22: 20976361-20976361 on Build GRCh38
rs753935603 Chr.22: 20976334-20976334 on Build GRCh38
rs1049345982 Chr.22: 20976420-20976420 on Build GRCh38
rs572459854 Chr.22: 20976277-20976277 on Build GRCh38
rs781439692 Chr.22: 20976560-20976560 on Build GRCh38
rs377242174 Chr.22: 20976382-20976382 on Build GRCh38
rs376685162 Chr.22: 20976179-20976179 on Build GRCh38
rs201385474 Chr.22: 20976333-20976333 on Build GRCh38
rs767779739 Chr.22: 20976198-20976198 on Build GRCh38
rs557504704 Chr.22: 20976391-20976391 on Build GRCh38
rs774658687 Chr.22: 20976312-20976312 on Build GRCh38
rs756384777 Chr.22: 20976284-20976284 on Build GRCh38

View Full Product Details