The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309193.1 | NP_001296122.1 | 7 - 8 | 1736 | 63 |
NM_004442.7 | NP_004433.2 | 7 - 8 | 1736 | 63 | |
NM_017449.4 | NP_059145.2 | 7 - 8 | 1736 | 63 | |
XM_006710441.3 | XP_006710504.1 | 7 - 8 | 1829 | 63 | |
XM_006710442.3 | XP_006710505.1 | 7 - 8 | 1711 | 63 | |
GenBank mRNA | AB210018.1 | - | 7 - 8 | 1645 | 63 |
AF025304.1 | - | 7 - 8 | 1616 | 63 | |
AK298898.1 | - | 3 - 4 | 413 | 63 | |
BC007903.2 | - | 6 - 7 | 1514 | 63 | |
BC018763.1 | - | 5 - 6 | 889 | 63 | |
D37827.1 | - | 1 - 2 | 67 | 63 | |
L36643.1 | - | 7 - 8 | 1543 | 63 | |
L41939.1 | - | 7 - 8 | 1695 | 63 |