The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005798.4 | NP_005789.2 | 2 - 3 | 222 | 71 |
NM_052811.3 | NP_434698.1 | 2 - 3 | 307 | 71 | |
NM_213590.2 | NP_998755.1 | 1 - 2 | 757 | 71 | |
GenBank mRNA | AF220127.1 | - | 2 - 3 | 162 | 71 |
AF241850.1 | - | 2 - 3 | 180 | 71 | |
AK314496.1 | - | 2 - 3 | 266 | 71 | |
AL360196.1 | - | 2 - 3 | 162 | 71 | |
AY191002.1 | - | 2 - 3 | 255 | 71 | |
AY455758.1 | - | 1 - 2 | 732 | 71 | |
BC003579.1 | - | 2 - 3 | 268 | 71 | |
BC063407.1 | - | 1 - 2 | 699 | 71 |