The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005868.5 | NP_005859.1 | 3 - 4 | 371 | 118 |
GenBank mRNA | AF007551.1 | - | 3 - 4 | 324 | 118 |
AK313984.1 | - | 3 - 4 | 344 | 118 | |
BC000899.1 | - | 3 - 4 | 260 | 118 | |
BT009853.1 | - | 3 - 4 | 205 | 118 |