Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50482886-50483375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM2
Entrez Gene ID
79924
Gene Name
adrenomedullin 2
Gene Aliases
AM2, dJ579N16.4
UniGene
Hs.449099
Gene Chromosome Location
Chr.22: 50481724-50486440 on Build GRCh38
Transcripts
NM_001253845.1 XM_017028940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7249364 c.509G>A p.G170D Chr.22: 50483215-50483215 on Build GRCh38
3842957 c.566G>A p.R189K Chr.22: 50483272-50483272 on Build GRCh38
5474464 c.366G>A p.T122T Chr.22: 50483072-50483072 on Build GRCh38
6595253 c.198G>T p.E66D Chr.22: 50482904-50482904 on Build GRCh38
3842956 c.559C>T p.L187L Chr.22: 50483265-50483265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009120730 Chr.22: 50483282-50483282 on Build GRCh38
rs903628177 Chr.22: 50483243-50483243 on Build GRCh38
rs775038143 Chr.22: 50483037-50483037 on Build GRCh38
rs746081346 Chr.22: 50482951-50482951 on Build GRCh38
rs766321313 Chr.22: 50482908-50482908 on Build GRCh38
rs565290435 Chr.22: 50483318-50483318 on Build GRCh38
rs61732427 Chr.22: 50482954-50482954 on Build GRCh38
rs548585514 Chr.22: 50483321-50483321 on Build GRCh38
rs561123799 Chr.22: 50483286-50483286 on Build GRCh38
rs956362577 Chr.22: 50483279-50483279 on Build GRCh38
rs999672474 Chr.22: 50483257-50483257 on Build GRCh38
rs560281953 Chr.22: 50483038-50483038 on Build GRCh38
rs911894035 Chr.22: 50483028-50483028 on Build GRCh38
rs532042958 Chr.22: 50483312-50483312 on Build GRCh38
rs2236031 Chr.22: 50483265-50483265 on Build GRCh38
rs112907082 Chr.22: 50483168-50483168 on Build GRCh38
rs61732429 Chr.22: 50483289-50483289 on Build GRCh38
rs754170812 Chr.22: 50483064-50483064 on Build GRCh38
rs770576228 Chr.22: 50482997-50482997 on Build GRCh38
rs939259391 Chr.22: 50483191-50483191 on Build GRCh38
rs764618296 Chr.22: 50483047-50483047 on Build GRCh38
rs988542320 Chr.22: 50483116-50483116 on Build GRCh38
rs531387314 Chr.22: 50483254-50483254 on Build GRCh38
rs764410445 Chr.22: 50482928-50482928 on Build GRCh38
rs924122165 Chr.22: 50483039-50483039 on Build GRCh38
rs974738034 Chr.22: 50483302-50483302 on Build GRCh38
rs1043401694 Chr.22: 50483237-50483237 on Build GRCh38
rs780311655 Chr.22: 50483126-50483126 on Build GRCh38
rs1011892189 Chr.22: 50483228-50483228 on Build GRCh38
rs574891822 Chr.22: 50483149-50483149 on Build GRCh38
rs1031128059 Chr.22: 50483244-50483244 on Build GRCh38
rs373496680 Chr.22: 50483330-50483330 on Build GRCh38
rs577386266 Chr.22: 50483080-50483080 on Build GRCh38
rs956896559 Chr.22: 50483260-50483260 on Build GRCh38
rs956348244 Chr.22: 50483015-50483015 on Build GRCh38
rs537686058 Chr.22: 50482992-50482992 on Build GRCh38
rs888545527 Chr.22: 50483196-50483196 on Build GRCh38
rs930027501 Chr.22: 50483175-50483175 on Build GRCh38
rs751493325 Chr.22: 50483285-50483285 on Build GRCh38
rs768112003 Chr.22: 50482911-50482922 on Build GRCh38
rs541555248 Chr.22: 50483085-50483085 on Build GRCh38
rs779222469 Chr.22: 50482947-50482947 on Build GRCh38
rs61732428 Chr.22: 50483313-50483313 on Build GRCh38
rs987647903 Chr.22: 50483017-50483017 on Build GRCh38
rs757445676 Chr.22: 50482941-50482941 on Build GRCh38
rs765680187 Chr.22: 50483097-50483097 on Build GRCh38
rs766300862 Chr.22: 50483305-50483305 on Build GRCh38
rs1040885840 Chr.22: 50482934-50482934 on Build GRCh38
rs758659354 Chr.22: 50483117-50483117 on Build GRCh38
rs762811921 Chr.22: 50483227-50483227 on Build GRCh38
rs767065117 Chr.22: 50483034-50483034 on Build GRCh38
rs1048276742 Chr.22: 50483338-50483338 on Build GRCh38
rs756622969 Chr.22: 50482926-50482926 on Build GRCh38
rs1000400122 Chr.22: 50482969-50482969 on Build GRCh38
rs1049421948 Chr.22: 50483137-50483137 on Build GRCh38
rs756082631 Chr.22: 50482986-50482986 on Build GRCh38
rs911090470 Chr.22: 50483139-50483139 on Build GRCh38
rs754194801 Chr.22: 50482931-50482931 on Build GRCh38
rs750720500 Chr.22: 50483099-50483099 on Build GRCh38
rs776750250 Chr.22: 50483148-50483148 on Build GRCh38
rs377126873 Chr.22: 50483036-50483036 on Build GRCh38
rs955784320 Chr.22: 50483050-50483050 on Build GRCh38
rs1026984558 Chr.22: 50482978-50482978 on Build GRCh38
rs1012638657 Chr.22: 50483210-50483210 on Build GRCh38
rs759054465 Chr.22: 50482975-50482975 on Build GRCh38
rs769476494 Chr.22: 50483049-50483049 on Build GRCh38
rs969307239 Chr.22: 50482957-50482957 on Build GRCh38
rs756019768 Chr.22: 50483164-50483164 on Build GRCh38
rs767012085 Chr.22: 50482912-50482912 on Build GRCh38
rs770770624 Chr.22: 50483225-50483225 on Build GRCh38
rs769729265 Chr.22: 50482955-50482955 on Build GRCh38
rs758606241 Chr.22: 50482953-50482953 on Build GRCh38
rs754100884 Chr.22: 50483353-50483353 on Build GRCh38
rs1019489500 Chr.22: 50482906-50482906 on Build GRCh38
rs971022877 Chr.22: 50483029-50483029 on Build GRCh38
rs759177930 Chr.22: 50483016-50483016 on Build GRCh38
rs559746568 Chr.22: 50483144-50483144 on Build GRCh38
rs748543119 Chr.22: 50483115-50483116 on Build GRCh38
rs766563384 Chr.22: 50483346-50483346 on Build GRCh38
rs897029059 Chr.22: 50483169-50483169 on Build GRCh38
rs753261161 Chr.22: 50482916-50482916 on Build GRCh38
rs889720378 Chr.22: 50482927-50482927 on Build GRCh38
rs774057338 Chr.22: 50483006-50483006 on Build GRCh38
rs529687790 Chr.22: 50483320-50483320 on Build GRCh38
rs929767567 Chr.22: 50483326-50483326 on Build GRCh38
rs918411630 Chr.22: 50483325-50483325 on Build GRCh38
rs777249467 Chr.22: 50483292-50483292 on Build GRCh38
rs13056677 Chr.22: 50483215-50483215 on Build GRCh38

View Full Product Details