The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037174.1 | NP_001032251.1 | 3 - 4 | 320 | 128 |
NM_012097.3 | NP_036229.1 | 3 - 4 | 576 | 128 | |
NM_177985.2 | NP_817114.2 | 3 - 4 | 318 | 128 | |
GenBank mRNA | AF087889.1 | - | - | 506 | 128 |
AF100740.1 | - | 3 - 4 | 323 | 128 | |
AF493891.1 | - | 3 - 4 | 257 | 128 | |
AK292714.1 | - | 3 - 4 | 486 | 128 | |
AK298937.1 | - | 3 - 4 | 264 | 128 | |
BC001254.1 | - | 3 - 4 | 494 | 128 | |
BG707385.1 | - | 3 - 4 | 297 | 128 | |
BT009829.1 | - | 3 - 4 | 257 | 128 | |
BX461356.2 | - | 3 - 4 | 291 | 128 | |
CX867116.1 | - | 3 - 4 | 582 | 128 |