Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
Chromosome Location
Chr.1: 151510859 - 151538692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020770.2 NP_065821.1 9 - 10 1904 91
XM_005245365.4 XP_005245422.1 9 - 10 1916 91
GenBank mRNA AB037740.1 - 9 - 10 1877 91
AF263462.1 - 9 - 10 1911 91
AK290007.1 - 9 - 10 1877 91
BC146657.1 - 9 - 10 1833 91
BC150604.1 - 8 - 9 1783 91
BC152445.1 - 9 - 10 1877 91

View Full Product Details