The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020919.3 | NP_065970.2 | 14 - 15 | 3065 | 62 |
XM_006712654.2 | XP_006712717.1 | 14 - 15 | 3642 | 62 | |
XM_006712655.3 | XP_006712718.1 | 10 - 11 | 1279 | 62 | |
XM_011511530.2 | XP_011509832.1 | 13 - 14 | 2777 | 62 | |
XM_017004569.1 | XP_016860058.1 | 13 - 14 | 2769 | 62 | |
XM_017004570.1 | XP_016860059.1 | 13 - 14 | 2769 | 62 | |
XM_017004571.1 | XP_016860060.1 | 13 - 14 | 2707 | 62 | |
XM_017004572.1 | XP_016860061.1 | 11 - 12 | 2357 | 62 | |
GenBank mRNA | AB046783.2 | - | 13 - 14 | 2786 | 62 |
AB053305.1 | - | 14 - 15 | 2832 | 62 | |
AF391100.1 | - | 14 - 15 | 2832 | 62 | |
AK056413.1 | - | 5 - 6 | 670 | 62 | |
AK291029.1 | - | 14 - 15 | 2849 | 62 | |
BC071576.1 | - | 14 - 15 | 2827 | 62 |