The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048164.2 | NP_001041629.1 | 3 - 4 | 679 | 64 |
NM_032803.5 | NP_116192.4 | 3 - 4 | 703 | 64 | |
XM_017029912.1 | XP_016885401.1 | 2 - 3 | 1194 | 64 | |
GenBank mRNA | AF320612.1 | - | 3 - 4 | 654 | 64 |
AK074865.1 | - | 3 - 4 | 674 | 64 | |
AK075014.1 | - | 3 - 4 | 679 | 64 | |
BC033816.1 | - | 3 - 4 | 658 | 64 |