Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORO6
Entrez Gene ID
84940
Gene Name
coronin 6
Gene Aliases
-
Chromosome Location
Chr.17: 29614756 - 29622912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005258056.4 XP_005258113.1 5 - 6 1397 124
XM_011525385.2 XP_011523687.1 10 - 11 1517 124
GenBank mRNA AF193039.1 - 4 - 5 499 124
AF447884.1 - 6 - 7 1282 124
AF447885.1 - 4 - 5 968 124
AK027777.1 - 4 - 5 970 124
AK092430.1 - 7 - 8 1213 124
BC048209.1 - 6 - 7 712 124
BC064514.1 - 6 - 7 712 124

View Full Product Details