The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271022.1 | NP_001257951.1 | 12 - 13 | 1974 | 112 |
NM_001271023.1 | NP_001257952.1 | 11 - 12 | 2231 | 112 | |
NM_130384.2 | NP_569055.1 | 11 - 12 | 2184 | 112 | |
GenBank mRNA | AF451323.1 | - | 11 - 12 | 2080 | 112 |
AK300548.1 | - | 12 - 13 | 1947 | 112 | |
AK315075.1 | - | 11 - 12 | 2148 | 112 |