Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73997856-73998384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL5
Entrez Gene ID
6374
Gene Name
C-X-C motif chemokine ligand 5
Gene Aliases
ENA-78, SCYB5
UniGene
Hs.89714
Gene Chromosome Location
Chr.4: 73995642-73998729 on Build GRCh38
Transcripts
NM_002994.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7081091 c.252G>A p.L84L Chr.4: 73998086-73998086 on Build GRCh38
1057479 c.324C>T p.D108D Chr.4: 73998014-73998014 on Build GRCh38
6638450 c.142C>T p.R48C Chr.4: 73998306-73998306 on Build GRCh38
6638451 c.177T>C p.H59H Chr.4: 73998271-73998271 on Build GRCh38
6545519 c.179C>A p.P60H Chr.4: 73998269-73998269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054904193 Chr.4: 73998093-73998093 on Build GRCh38
rs200635427 Chr.4: 73998240-73998240 on Build GRCh38
rs904790507 Chr.4: 73998358-73998358 on Build GRCh38
rs146439766 Chr.4: 73998127-73998127 on Build GRCh38
rs780470589 Chr.4: 73998046-73998046 on Build GRCh38
rs752604189 Chr.4: 73998184-73998184 on Build GRCh38
rs425535 Chr.4: 73998280-73998280 on Build GRCh38
rs757406987 Chr.4: 73998209-73998209 on Build GRCh38
rs1014271028 Chr.4: 73998020-73998020 on Build GRCh38
rs200193764 Chr.4: 73998004-73998004 on Build GRCh38
rs753834662 Chr.4: 73998017-73998017 on Build GRCh38
rs754782309 Chr.4: 73998024-73998024 on Build GRCh38
rs904569505 Chr.4: 73997960-73997960 on Build GRCh38
rs746630157 Chr.4: 73998339-73998339 on Build GRCh38
rs775817309 Chr.4: 73997986-73997986 on Build GRCh38
rs369811933 Chr.4: 73998231-73998231 on Build GRCh38
rs201162216 Chr.4: 73997964-73997964 on Build GRCh38
rs569405414 Chr.4: 73997931-73997931 on Build GRCh38
rs201976415 Chr.4: 73998340-73998340 on Build GRCh38
rs775301113 Chr.4: 73998259-73998259 on Build GRCh38
rs1016006155 Chr.4: 73997996-73997996 on Build GRCh38
rs200088477 Chr.4: 73998346-73998346 on Build GRCh38
rs764910847 Chr.4: 73998141-73998141 on Build GRCh38
rs201793265 Chr.4: 73998052-73998052 on Build GRCh38
rs753178513 Chr.4: 73998316-73998316 on Build GRCh38
rs770545074 Chr.4: 73998249-73998249 on Build GRCh38
rs750647172 Chr.4: 73998223-73998223 on Build GRCh38
rs952391441 Chr.4: 73998183-73998183 on Build GRCh38
rs534636782 Chr.4: 73998028-73998028 on Build GRCh38
rs776321799 Chr.4: 73998252-73998252 on Build GRCh38
rs901311019 Chr.4: 73997881-73997881 on Build GRCh38
rs200648901 Chr.4: 73998338-73998338 on Build GRCh38
rs200608132 Chr.4: 73998269-73998269 on Build GRCh38
rs193295351 Chr.4: 73998327-73998327 on Build GRCh38
rs770775855 Chr.4: 73998343-73998343 on Build GRCh38
rs563521523 Chr.4: 73998279-73998279 on Build GRCh38
rs773832107 Chr.4: 73997999-73997999 on Build GRCh38
rs201878647 Chr.4: 73998116-73998116 on Build GRCh38
rs368292646 Chr.4: 73997992-73997992 on Build GRCh38
rs545420380 Chr.4: 73998084-73998084 on Build GRCh38
rs879183368 Chr.4: 73998205-73998205 on Build GRCh38
rs543588409 Chr.4: 73998283-73998283 on Build GRCh38
rs142384343 Chr.4: 73998360-73998360 on Build GRCh38
rs769104652 Chr.4: 73997988-73997991 on Build GRCh38
rs779413033 Chr.4: 73998306-73998306 on Build GRCh38
rs371956029 Chr.4: 73998005-73998005 on Build GRCh38
rs751708366 Chr.4: 73998031-73998031 on Build GRCh38
rs200900311 Chr.4: 73998359-73998359 on Build GRCh38
rs759595586 Chr.4: 73998131-73998131 on Build GRCh38
rs759542018 Chr.4: 73998254-73998254 on Build GRCh38
rs1041627589 Chr.4: 73997879-73997879 on Build GRCh38
rs111813861 Chr.4: 73998013-73998013 on Build GRCh38
rs774823743 Chr.4: 73998077-73998077 on Build GRCh38
rs369381841 Chr.4: 73998009-73998009 on Build GRCh38
rs199818484 Chr.4: 73998312-73998312 on Build GRCh38
rs545218332 Chr.4: 73998268-73998268 on Build GRCh38
rs756143598 Chr.4: 73998224-73998224 on Build GRCh38
rs376176437 Chr.4: 73998007-73998007 on Build GRCh38
rs199676653 Chr.4: 73998233-73998233 on Build GRCh38
rs758741757 Chr.4: 73998328-73998328 on Build GRCh38
rs771818240 Chr.4: 73998089-73998089 on Build GRCh38
rs551599111 Chr.4: 73997909-73997909 on Build GRCh38
rs187709912 Chr.4: 73998301-73998301 on Build GRCh38
rs970576965 Chr.4: 73998092-73998092 on Build GRCh38
rs373741602 Chr.4: 73998096-73998096 on Build GRCh38
rs375096397 Chr.4: 73998120-73998120 on Build GRCh38
rs746016952 Chr.4: 73998043-73998043 on Build GRCh38
rs748488678 Chr.4: 73998345-73998348 on Build GRCh38
rs886388735 Chr.4: 73998310-73998311 on Build GRCh38
rs748490975 Chr.4: 73998235-73998235 on Build GRCh38
rs781737431 Chr.4: 73998130-73998131 on Build GRCh38
rs573521158 Chr.4: 73998014-73998014 on Build GRCh38
rs781126126 Chr.4: 73998041-73998041 on Build GRCh38
rs767718489 Chr.4: 73998216-73998216 on Build GRCh38
rs536772191 Chr.4: 73997944-73997944 on Build GRCh38
rs997404230 Chr.4: 73997921-73997921 on Build GRCh38
rs150702442 Chr.4: 73998304-73998304 on Build GRCh38
rs200220379 Chr.4: 73998227-73998227 on Build GRCh38
rs983797283 Chr.4: 73998203-73998203 on Build GRCh38
rs879005377 Chr.4: 73998204-73998205 on Build GRCh38
rs772330492 Chr.4: 73998237-73998237 on Build GRCh38
rs984889752 Chr.4: 73997984-73997984 on Build GRCh38
rs756196974 Chr.4: 73998045-73998045 on Build GRCh38
rs772756603 Chr.4: 73998094-73998094 on Build GRCh38
rs200254576 Chr.4: 73998053-73998053 on Build GRCh38
rs981120666 Chr.4: 73998098-73998098 on Build GRCh38
rs747755561 Chr.4: 73998083-73998083 on Build GRCh38
rs765018221 Chr.4: 73998035-73998035 on Build GRCh38
rs3864158 Chr.4: 73998138-73998138 on Build GRCh38
rs199518894 Chr.4: 73998263-73998263 on Build GRCh38
rs1016312527 Chr.4: 73998324-73998324 on Build GRCh38
rs201814566 Chr.4: 73997989-73997989 on Build GRCh38
rs148775944 Chr.4: 73998152-73998152 on Build GRCh38
rs200883747 Chr.4: 73998074-73998074 on Build GRCh38
rs1017816088 Chr.4: 73997888-73997888 on Build GRCh38
rs201539502 Chr.4: 73998230-73998230 on Build GRCh38
rs201283785 Chr.4: 73998255-73998255 on Build GRCh38

View Full Product Details