Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCD
Entrez Gene ID
117159
Gene Name
dermcidin
Gene Aliases
AIDD, DCD-1, DSEP, HCAP, PIF
Chromosome Location
Chr.12: 54644591 - 54648493 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300854.1 NP_001287783.1 1 - 2 247 120
NM_053283.3 NP_444513.1 1 - 2 247 120
GenBank mRNA AF144011.1 - 1 - 2 117 120
AY590150.1 - 1 - 2 111 120
BC062682.1 - 1 - 2 119 120
BC069108.1 - 1 - 2 117 120
BP313072.1 - 1 - 2 119 120
EF503687.1 - 1 - 2 247 120
EF503688.1 - 1 - 2 247 287
EF503689.1 - 1 - 2 247 120

View Full Product Details