The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282309.1 | NP_001269238.1 | 8 - 9 | 955 | 64 |
NM_001282310.1 | NP_001269239.1 | 7 - 8 | 870 | 64 | |
NM_054020.3 | NP_473361.1 | 7 - 8 | 1056 | 64 | |
NM_172095.2 | NP_742093.1 | 7 - 8 | 1056 | 64 | |
NR_110319.1 | 6 - 7 | 843 | 64 | ||
XM_017021904.1 | XP_016877393.1 | 7 - 8 | 1062 | 64 | |
GenBank mRNA | AF411817.1 | - | 6 - 7 | 841 | 64 |
AF411818.1 | - | 6 - 7 | 841 | 64 | |
AF411819.1 | - | 6 - 7 | 841 | 64 | |
BC064387.1 | - | 7 - 8 | 870 | 64 | |
BX648099.1 | - | 7 - 8 | 1092 | 64 |