The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057161.3 | NP_476502.1 | 10 - 11 | 1382 | 79 |
NR_040101.1 | 9 - 10 | 1266 | 79 | ||
GenBank mRNA | AB055925.1 | - | 10 - 11 | 1246 | 79 |
AK097719.1 | - | 8 - 9 | 1108 | 79 | |
AK290384.1 | - | 10 - 11 | 1238 | 79 | |
AK302336.1 | - | 9 - 10 | 1021 | 79 | |
BC000295.2 | - | 10 - 11 | 1202 | 79 | |
BC001789.1 | - | 10 - 11 | 1217 | 79 | |
BC001793.2 | - | 10 - 11 | 1180 | 79 | |
BC007296.1 | - | 10 - 11 | 1232 | 79 | |
BC009460.1 | - | 10 - 11 | 1230 | 79 | |
BC021546.1 | - | 10 - 11 | 1219 | 79 | |
BC041793.1 | - | 9 - 10 | 1120 | 79 | |
BC045612.1 | - | 9 - 10 | 1120 | 79 |