The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078.3 | NP_001069.1 | 1 - 2 | 280 | 109 |
NM_001199834.1 | NP_001186763.1 | 1 - 2 | 280 | 109 | |
NM_080682.2 | NP_542413.1 | 1 - 2 | 280 | 109 | |
GenBank mRNA | AK223266.1 | - | 1 - 2 | 179 | 109 |
AK291732.1 | - | 1 - 2 | 178 | 109 | |
AK296692.1 | - | 1 - 2 | 178 | 109 | |
AU121762.1 | - | 1 - 2 | 180 | 109 | |
BC017276.2 | - | 1 - 2 | 160 | 109 | |
BC068490.1 | - | 1 - 2 | 178 | 109 | |
BC085003.1 | - | 1 - 2 | 160 | 109 | |
DB096825.1 | - | 1 - 2 | 280 | 109 | |
M30257.1 | - | 1 - 2 | 165 | 109 | |
M60335.1 | - | 1 - 2 | 158 | 109 | |
X53051.1 | - | 1 - 2 | 59 | 109 |