Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMELX
Entrez Gene ID
265
Gene Name
amelogenin, X-linked
Gene Aliases
AI1E, AIH1, ALGN, AMG, AMGL, AMGX
Chromosome Location
Chr.X: 11293413 - 11300761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142.2 NP_001133.1 1 - 2 58 91
NM_182680.1 NP_872621.1 1 - 2 58 91
NM_182681.1 NP_872622.1 1 - 2 58 91
GenBank mRNA AF436849.1 - 1 - 2 58 91
BC069118.1 - 1 - 2 58 91
M86932.1 - 1 - 2 58 91

View Full Product Details