The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033044.3 | NP_001028216.1 | 2 - 3 | 711 | 81 |
NM_001033056.3 | NP_001028228.1 | 2 - 3 | 413 | 81 | |
NM_002065.6 | NP_002056.2 | 3 - 4 | 1067 | 81 | |
XM_006711278.1 | XP_006711341.1 | 2 - 3 | 774 | 81 | |
GenBank mRNA | AK092874.1 | - | 1 - 2 | 1731 | 81 |
AK122784.1 | - | 2 - 3 | 711 | 81 | |
AL161952.1 | - | 1 - 2 | 600 | 81 | |
AY486122.1 | - | 1 - 2 | 170 | 81 | |
AY513283.1 | - | 2 - 3 | 226 | 81 | |
AY947343.1 | - | 2 - 3 | 271 | 81 | |
BC010037.2 | - | 2 - 3 | 283 | 81 | |
BC011700.2 | - | 2 - 3 | 279 | 81 | |
BC011852.2 | - | 2 - 3 | 290 | 81 | |
BC018992.1 | - | 3 - 4 | 630 | 81 | |
BC031964.1 | - | 2 - 3 | 813 | 81 | |
BC051726.1 | - | 2 - 3 | 1476 | 81 | |
BC127883.1 | - | 1 - 2 | 81 | 81 | |
BG771732.1 | - | 2 - 3 | 330 | 81 | |
BX537384.1 | - | 2 - 3 | 295 | 81 | |
CD688156.1 | - | 2 - 3 | 404 | 81 | |
DA151535.1 | - | 2 - 3 | 356 | 81 | |
DA456371.1 | - | 2 - 3 | 414 | 81 | |
S70290.1 | - | 1 - 2 | 170 | 81 | |
X59834.1 | - | 2 - 3 | 279 | 81 | |
Y00387.2 | - | 2 - 3 | 289 | 81 |