Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr.11: 47269376 - 47330031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135943.1 NP_001129415.1 6 - 7 1424 79
NM_001135944.1 NP_001129416.1 6 - 7 1400 79
NM_003682.3 NP_003673.3 6 - 7 1400 79
NM_130470.2 NP_569826.2 6 - 7 1566 79
NM_130471.2 NP_569827.2 6 - 7 1400 79
NM_130472.2 NP_569828.2 6 - 7 1400 79
NM_130473.2 NP_569829.2 6 - 7 1400 79
NM_130474.2 NP_569830.2 6 - 7 1400 79
NM_130475.2 NP_569831.1 6 - 7 1424 79
NM_130476.2 NP_569832.2 6 - 7 1424 79
XM_005253189.2 XP_005253246.1 6 - 7 1424 79
XM_005253196.2 XP_005253253.1 6 - 7 1424 79
XM_005253199.2 XP_005253256.1 6 - 7 1424 79
XM_005253200.2 XP_005253257.1 6 - 7 1424 79
XM_005253201.2 XP_005253258.1 6 - 7 1424 79
XM_005253203.2 XP_005253260.1 6 - 7 1424 79
XM_005253204.2 XP_005253261.1 6 - 7 1424 79
XM_005253205.1 XP_005253262.1 6 - 7 1424 79
XM_017018478.1 XP_016873967.1 6 - 7 1424 79
XM_017018479.1 XP_016873968.1 6 - 7 1424 79
XM_017018480.1 XP_016873969.1 6 - 7 1424 79
XM_017018481.1 XP_016873970.1 6 - 7 1424 79
XM_017018482.1 XP_016873971.1 6 - 7 1424 79
XM_017018483.1 XP_016873972.1 6 - 7 1424 79
XM_017018484.1 XP_016873973.1 6 - 7 1424 79
XM_017018485.1 XP_016873974.1 6 - 7 1385 79
XM_017018486.1 XP_016873975.1 6 - 7 1424 79
XM_017018487.1 XP_016873976.1 6 - 7 1424 79
XM_017018488.1 XP_016873977.1 6 - 7 1424 79
XM_017018489.1 XP_016873978.1 6 - 7 1385 79
XM_017018490.1 XP_016873979.1 6 - 7 1411 79
XM_017018491.1 XP_016873980.1 6 - 7 1380 79
XM_017018492.1 XP_016873981.1 6 - 7 1424 79
XM_017018493.1 XP_016873982.1 6 - 7 1411 79
XM_017018494.1 XP_016873983.1 6 - 7 1424 79
XM_017018495.1 XP_016873984.1 6 - 7 1424 79
XM_017018496.1 XP_016873985.1 6 - 7 1380 79
XM_017018497.1 XP_016873986.1 6 - 7 1385 79
XM_017018498.1 XP_016873987.1 6 - 7 1424 79
XM_017018499.1 XP_016873988.1 6 - 7 1393 79
XM_017018500.1 XP_016873989.1 6 - 7 1424 79
XM_017018501.1 XP_016873990.1 6 - 7 1380 79
XM_017018502.1 XP_016873991.1 6 - 7 1393 79
XM_017018503.1 XP_016873992.1 6 - 7 1380 79
XM_017018504.1 XP_016873993.1 6 - 7 1393 79
XM_017018505.1 XP_016873994.1 6 - 7 1380 79
XM_017018506.1 XP_016873995.1 6 - 7 1385 79
XM_017018507.1 XP_016873996.1 6 - 7 1411 79
XM_017018508.1 XP_016873997.1 6 - 7 1424 79
XM_017018509.1 XP_016873998.1 6 - 7 1380 79
XM_017018510.1 XP_016873999.1 6 - 7 1424 79
GenBank mRNA AB002356.1 - 6 - 7 1396 79
AF440100.1 - 6 - 7 1374 79
AF440101.1 - 6 - 7 1374 79
AF440102.1 - 6 - 7 1374 79
AF440103.1 - 6 - 7 1374 79
AF440434.1 - 6 - 7 1374 79
AK292442.1 - 6 - 7 1364 79
BC040484.1 - 6 - 7 1408 79
U44953.1 - 6 - 7 1388 79
U77352.1 - 6 - 7 1534 79

View Full Product Details