Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr.11: 47269376 - 47330031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135943.1 NP_001129415.1 20 - 21 3484 66
NM_001135944.1 NP_001129416.1 20 - 21 3460 66
NM_003682.3 NP_003673.3 22 - 23 3703 66
NM_130470.2 NP_569826.2 20 - 21 3755 66
NM_130471.2 NP_569827.2 21 - 22 3520 66
NM_130472.2 NP_569828.2 20 - 21 3460 66
NM_130473.2 NP_569829.2 21 - 22 3649 66
NM_130474.2 NP_569830.2 20 - 21 3460 66
NM_130475.2 NP_569831.1 22 - 23 3727 66
NM_130476.2 NP_569832.2 20 - 21 3613 66
XM_005253189.2 XP_005253246.1 22 - 23 3727 66
XM_005253196.2 XP_005253253.1 21 - 22 3673 66
XM_005253199.2 XP_005253256.1 20 - 21 3613 66
XM_005253200.2 XP_005253257.1 22 - 23 3727 66
XM_005253201.2 XP_005253258.1 21 - 22 3544 66
XM_005253203.2 XP_005253260.1 20 - 21 3484 66
XM_005253204.2 XP_005253261.1 20 - 21 3484 66
XM_005253205.1 XP_005253262.1 20 - 21 3484 66
XM_017018478.1 XP_016873967.1 22 - 23 3727 66
XM_017018479.1 XP_016873968.1 21 - 22 3673 66
XM_017018480.1 XP_016873969.1 21 - 22 3673 66
XM_017018481.1 XP_016873970.1 20 - 21 3613 66
XM_017018482.1 XP_016873971.1 21 - 22 3673 66
XM_017018483.1 XP_016873972.1 22 - 23 3598 66
XM_017018484.1 XP_016873973.1 20 - 21 3613 66
XM_017018485.1 XP_016873974.1 20 - 21 3574 66
XM_017018486.1 XP_016873975.1 21 - 22 3544 66
XM_017018487.1 XP_016873976.1 21 - 22 3538 66
XM_017018488.1 XP_016873977.1 22 - 23 3727 66
XM_017018489.1 XP_016873978.1 22 - 23 3688 66
XM_017018490.1 XP_016873979.1 21 - 22 3660 66
XM_017018491.1 XP_016873980.1 21 - 22 3629 66
XM_017018492.1 XP_016873981.1 21 - 22 3673 66
XM_017018493.1 XP_016873982.1 21 - 22 3654 66
XM_017018494.1 XP_016873983.1 20 - 21 3613 66
XM_017018495.1 XP_016873984.1 20 - 21 3613 66
XM_017018496.1 XP_016873985.1 20 - 21 3569 66
XM_017018497.1 XP_016873986.1 20 - 21 3574 66
XM_017018498.1 XP_016873987.1 22 - 23 3598 66
XM_017018499.1 XP_016873988.1 22 - 23 3567 66
XM_017018500.1 XP_016873989.1 20 - 21 3613 66
XM_017018501.1 XP_016873990.1 21 - 22 3500 66
XM_017018502.1 XP_016873991.1 21 - 22 3513 66
XM_017018503.1 XP_016873992.1 21 - 22 3500 66
XM_017018504.1 XP_016873993.1 21 - 22 3513 66
XM_017018505.1 XP_016873994.1 21 - 22 3500 66
XM_017018506.1 XP_016873995.1 21 - 22 3505 66
XM_017018507.1 XP_016873996.1 21 - 22 3525 66
XM_017018508.1 XP_016873997.1 20 - 21 3484 66
XM_017018509.1 XP_016873998.1 20 - 21 3440 66
XM_017018510.1 XP_016873999.1 20 - 21 3484 66
GenBank mRNA AB002356.1 - 22 - 23 3699 66
AF440100.1 - 22 - 23 3677 66
AF440101.1 - 21 - 22 3623 66
AF440102.1 - 21 - 22 3494 66
AF440103.1 - 20 - 21 3434 66
AF440434.1 - 20 - 21 3434 66
AK292442.1 - 20 - 21 3424 66
AK308780.1 - 3 - 4 167 66
BC040484.1 - 20 - 21 3468 66
U44953.1 - 20 - 21 3577 66
U77352.1 - 20 - 21 3723 66

View Full Product Details