Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr.11: 47269376 - 47330031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135943.1 NP_001129415.1 26 - 27 4174 81
NM_001135944.1 NP_001129416.1 26 - 27 4141 81
NM_003682.3 NP_003673.3 29 - 30 4459 81
NM_130470.2 NP_569826.2 26 - 27 4448 81
NM_130471.2 NP_569827.2 27 - 28 4213 81
NM_130472.2 NP_569828.2 26 - 27 4153 81
NM_130473.2 NP_569829.2 27 - 28 4342 81
NM_130474.2 NP_569830.2 26 - 27 4153 81
NM_130475.2 NP_569831.1 29 - 30 4483 81
NM_130476.2 NP_569832.2 26 - 27 4303 81
XM_005253189.2 XP_005253246.1 29 - 30 4483 81
XM_005253196.2 XP_005253253.1 27 - 28 4366 81
XM_005253199.2 XP_005253256.1 26 - 27 4306 81
XM_005253200.2 XP_005253257.1 29 - 30 4480 81
XM_005253201.2 XP_005253258.1 27 - 28 4237 81
XM_005253203.2 XP_005253260.1 26 - 27 4177 81
XM_005253204.2 XP_005253261.1 26 - 27 4165 81
XM_005253205.1 XP_005253262.1 26 - 27 4177 81
XM_017018478.1 XP_016873967.1 29 - 30 4471 81
XM_017018479.1 XP_016873968.1 28 - 29 4417 81
XM_017018480.1 XP_016873969.1 27 - 28 4363 81
XM_017018481.1 XP_016873970.1 27 - 28 4357 81
XM_017018482.1 XP_016873971.1 27 - 28 4354 81
XM_017018483.1 XP_016873972.1 29 - 30 4342 81
XM_017018484.1 XP_016873973.1 26 - 27 4294 81
XM_017018485.1 XP_016873974.1 26 - 27 4255 81
XM_017018486.1 XP_016873975.1 28 - 29 4288 81
XM_017018487.1 XP_016873976.1 28 - 29 4282 81
XM_017018488.1 XP_016873977.1 29 - 30 4471 81
XM_017018489.1 XP_016873978.1 29 - 30 4432 81
XM_017018490.1 XP_016873979.1 28 - 29 4416 81
XM_017018491.1 XP_016873980.1 28 - 29 4382 81
XM_017018492.1 XP_016873981.1 28 - 29 4417 81
XM_017018493.1 XP_016873982.1 28 - 29 4398 81
XM_017018494.1 XP_016873983.1 27 - 28 4369 81
XM_017018495.1 XP_016873984.1 27 - 28 4366 81
XM_017018496.1 XP_016873985.1 27 - 28 4313 81
XM_017018497.1 XP_016873986.1 27 - 28 4318 81
XM_017018498.1 XP_016873987.1 29 - 30 4342 81
XM_017018499.1 XP_016873988.1 29 - 30 4311 81
XM_017018500.1 XP_016873989.1 26 - 27 4303 81
XM_017018501.1 XP_016873990.1 28 - 29 4256 81
XM_017018502.1 XP_016873991.1 28 - 29 4269 81
XM_017018503.1 XP_016873992.1 28 - 29 4253 81
XM_017018504.1 XP_016873993.1 28 - 29 4266 81
XM_017018505.1 XP_016873994.1 28 - 29 4244 81
XM_017018506.1 XP_016873995.1 28 - 29 4249 81
XM_017018507.1 XP_016873996.1 28 - 29 4269 81
XM_017018508.1 XP_016873997.1 27 - 28 4240 81
XM_017018509.1 XP_016873998.1 27 - 28 4193 81
XM_017018510.1 XP_016873999.1 27 - 28 4228 81
GenBank mRNA AB002356.1 - 29 - 30 4455 81
AF440100.1 - 29 - 30 4433 81
AF440101.1 - 27 - 28 4316 81
AF440102.1 - 27 - 28 4187 81
AF440103.1 - 26 - 27 4127 81
AF440434.1 - 26 - 27 4127 81
AK292442.1 - 26 - 27 4105 81
AK308780.1 - 10 - 11 911 81
BC040484.1 - 26 - 27 4158 81
U44953.1 - 26 - 27 4267 81
U77352.1 - 26 - 27 4416 81

View Full Product Details