The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253908.1 | NP_001240837.1 | 8 - 9 | 978 | 112 |
NM_003739.5 | NP_003730.4 | 8 - 9 | 1001 | 112 | |
GenBank mRNA | AB018580.1 | - | 8 - 9 | 979 | 112 |
AK290365.1 | - | 8 - 9 | 961 | 112 | |
AK296829.1 | - | 8 - 9 | 1045 | 112 | |
BC001479.2 | - | 8 - 9 | 988 | 112 | |
BC019230.1 | - | 8 - 9 | 958 | 112 | |
D17793.1 | - | 8 - 9 | 983 | 112 |