Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NKG7
Entrez Gene ID
4818
Gene Name
natural killer cell granule protein 7
Gene Aliases
GIG1, GMP-17, p15-TIA-1
Chromosome Location
Chr.19: 51371616 - 51372706 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005601.3 NP_005592.1 2 - 3 471 81
XM_005258955.3 XP_005259012.1 2 - 3 348 81
GenBank mRNA AI313472.1 - 2 - 3 81
BC015759.1 - 2 - 3 400 81
S69115.1 - 2 - 3 480 81

View Full Product Details