The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001023567.4 | NP_001018861.3 | 7 - 8 | 678 | 63 |
NR_027410.1 | 15 - 16 | 2926 | 63 | ||
XM_011521596.1 | XP_011519898.1 | 14 - 15 | 1523 | 63 | |
XM_011521597.1 | XP_011519899.1 | 13 - 14 | 1474 | 63 | |
XM_017022210.1 | XP_016877699.1 | 15 - 16 | 1827 | 63 | |
XM_017022211.1 | XP_016877700.1 | 15 - 16 | 1596 | 63 | |
XM_017022212.1 | XP_016877701.1 | 14 - 15 | 2150 | 63 | |
XM_017022213.1 | XP_016877702.1 | 14 - 15 | 1552 | 63 | |
XM_017022214.1 | XP_016877703.1 | 15 - 16 | 1827 | 63 | |
XM_017022215.1 | XP_016877704.1 | 15 - 16 | 2349 | 63 | |
XM_017022216.1 | XP_016877705.1 | 14 - 15 | 2947 | 63 | |
XM_017022217.1 | XP_016877706.1 | 13 - 14 | 2869 | 63 | |
XM_017022218.1 | XP_016877707.1 | 15 - 16 | 2976 | 63 | |
GenBank mRNA | AK097551.1 | - | 7 - 8 | 667 | 63 |
BC143254.1 | - | 7 - 8 | 624 | 63 | |
BC144610.1 | - | 5 - 6 | 415 | 63 | |
BX647673.1 | - | 14 - 15 | 2895 | 63 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181077.3 | NP_851422.1 | 7 - 8 | 678 | 63 |
NR_027409.1 | 15 - 16 | 2940 | 63 | ||
XM_006720440.2 | XP_006720503.1 | 7 - 8 | 646 | 63 | |
XM_017022018.1 | XP_016877507.1 | 11 - 12 | 3074 | 63 | |
XM_017022019.1 | XP_016877508.1 | 10 - 11 | 2770 | 63 | |
XM_017022020.1 | XP_016877509.1 | 5 - 6 | 768 | 63 | |
GenBank mRNA | AB020662.1 | - | 6 - 7 | 658 | 63 |
AF204231.1 | - | 6 - 7 | 995 | 63 | |
AL137686.1 | - | 5 - 6 | 402 | 63 | |
BC032774.1 | - | 7 - 8 | 580 | 63 | |
BC093673.1 | - | 7 - 8 | 624 | 63 | |
BC104800.1 | - | 7 - 8 | 624 | 63 | |
BC144609.1 | - | 5 - 6 | 415 | 63 | |
BC150329.1 | - | 6 - 7 | 669 | 63 | |
BC152410.1 | - | 6 - 7 | 658 | 63 | |
BX648160.1 | - | 14 - 15 | 2894 | 63 | |
BX648575.1 | - | 14 - 15 | 2647 | 63 | |
CR749361.1 | - | 7 - 8 | 1831 | 63 |