Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTLL5
Entrez Gene ID
23093
Gene Name
tubulin tyrosine ligase like 5
Gene Aliases
CORD19, KIAA0998, STAMP
Chromosome Location
Chr.14: 75661208 - 75955082 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015072.4 NP_055887.3 23 - 24 2662 137
GenBank mRNA AB023215.1 - 21 - 22 2222 137
AK309748.1 - 24 - 25 2644 137
AL136808.1 - 9 - 10 1110 137
AY237126.1 - 23 - 24 2592 137
BC016625.1 - 10 - 11 1243 137
BC136472.1 - 23 - 24 2552 137
BC136473.1 - 23 - 24 2552 137
CR533554.1 - 8 - 9 1040 137

View Full Product Details