Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106512139-106512632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM1
Entrez Gene ID
202
Gene Name
absent in melanoma 1
Gene Aliases
CRYBG1, ST4
UniGene
Hs.724262
Gene Chromosome Location
Chr.6: 106360808-106570449 on Build GRCh38
Transcripts
NM_001624.3 XM_005266839.3 XM_017010333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6241085 c.112G>T p.A38S Chr.6: 106512453-106512453 on Build GRCh38
127286 c.144G>T p.A48A Chr.6: 106512485-106512485 on Build GRCh38
739788 c.189G>T p.R63R Chr.6: 106512530-106512530 on Build GRCh38
6028053 c.202delG p.G69fs*146 Chr.6: 106512543-106512543 on Build GRCh38
1238156 c.275C>G p.S92W Chr.6: 106512616-106512616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765002885 Chr.6: 106512486-106512486 on Build GRCh38
rs749367865 Chr.6: 106512576-106512576 on Build GRCh38
rs780857817 Chr.6: 106512328-106512328 on Build GRCh38
rs779534062 Chr.6: 106512461-106512463 on Build GRCh38
rs750280604 Chr.6: 106512493-106512493 on Build GRCh38
rs1013273101 Chr.6: 106512225-106512225 on Build GRCh38
rs749516587 Chr.6: 106512508-106512508 on Build GRCh38
rs771272419 Chr.6: 106512441-106512441 on Build GRCh38
rs374266954 Chr.6: 106512425-106512425 on Build GRCh38
rs113223052 Chr.6: 106512179-106512179 on Build GRCh38
rs939812492 Chr.6: 106512432-106512432 on Build GRCh38
rs754932179 Chr.6: 106512569-106512569 on Build GRCh38
rs746940343 Chr.6: 106512499-106512499 on Build GRCh38
rs752323326 Chr.6: 106512388-106512388 on Build GRCh38
rs773073604 Chr.6: 106512212-106512212 on Build GRCh38
rs1022077009 Chr.6: 106512258-106512258 on Build GRCh38
rs369540763 Chr.6: 106512360-106512360 on Build GRCh38
rs772589592 Chr.6: 106512529-106512529 on Build GRCh38
rs766947431 Chr.6: 106512310-106512310 on Build GRCh38
rs766687826 Chr.6: 106512376-106512376 on Build GRCh38
rs138380284 Chr.6: 106512442-106512442 on Build GRCh38
rs781241734 Chr.6: 106512414-106512414 on Build GRCh38
rs774826765 Chr.6: 106512525-106512525 on Build GRCh38
rs1010718510 Chr.6: 106512257-106512257 on Build GRCh38
rs760137463 Chr.6: 106512380-106512380 on Build GRCh38
rs760116953 Chr.6: 106512444-106512444 on Build GRCh38
rs771680430 Chr.6: 106512183-106512183 on Build GRCh38
rs956739001 Chr.6: 106512354-106512354 on Build GRCh38
rs756872369 Chr.6: 106512476-106512476 on Build GRCh38
rs1038104922 Chr.6: 106512440-106512440 on Build GRCh38
rs780012478 Chr.6: 106512498-106512498 on Build GRCh38
rs753943260 Chr.6: 106512447-106512461 on Build GRCh38
rs934133345 Chr.6: 106512170-106512170 on Build GRCh38
rs140169285 Chr.6: 106512558-106512558 on Build GRCh38
rs746285066 Chr.6: 106512526-106512526 on Build GRCh38
rs910019984 Chr.6: 106512366-106512366 on Build GRCh38
rs745834762 Chr.6: 106512353-106512353 on Build GRCh38
rs750663680 Chr.6: 106512164-106512169 on Build GRCh38
rs898182988 Chr.6: 106512446-106512446 on Build GRCh38
rs767257293 Chr.6: 106512161-106512162 on Build GRCh38
rs201831323 Chr.6: 106512364-106512364 on Build GRCh38
rs555538027 Chr.6: 106512363-106512363 on Build GRCh38
rs758138078 Chr.6: 106512561-106512561 on Build GRCh38
rs764179881 Chr.6: 106512259-106512259 on Build GRCh38
rs757531584 Chr.6: 106512587-106512587 on Build GRCh38
rs528778993 Chr.6: 106512197-106512197 on Build GRCh38
rs755889666 Chr.6: 106512329-106512329 on Build GRCh38
rs975974668 Chr.6: 106512597-106512597 on Build GRCh38
rs779999459 Chr.6: 106512382-106512382 on Build GRCh38
rs968472937 Chr.6: 106512568-106512568 on Build GRCh38
rs777426700 Chr.6: 106512333-106512333 on Build GRCh38
rs4945755 Chr.6: 106512164-106512164 on Build GRCh38
rs910072289 Chr.6: 106512590-106512590 on Build GRCh38
rs886889529 Chr.6: 106512290-106512290 on Build GRCh38
rs772589499 Chr.6: 106512504-106512505 on Build GRCh38
rs142945856 Chr.6: 106512465-106512465 on Build GRCh38
rs200417771 Chr.6: 106512458-106512458 on Build GRCh38
rs773415153 Chr.6: 106512371-106512371 on Build GRCh38
rs922400146 Chr.6: 106512612-106512612 on Build GRCh38
rs753454001 Chr.6: 106512402-106512402 on Build GRCh38
rs1039434211 Chr.6: 106512383-106512383 on Build GRCh38
rs755491880 Chr.6: 106512460-106512480 on Build GRCh38
rs751459745 Chr.6: 106512567-106512567 on Build GRCh38
rs761090045 Chr.6: 106512540-106512540 on Build GRCh38
rs572647174 Chr.6: 106512491-106512491 on Build GRCh38
rs11152999 Chr.6: 106512572-106512572 on Build GRCh38
rs775730176 Chr.6: 106512594-106512594 on Build GRCh38
rs892423159 Chr.6: 106512461-106512461 on Build GRCh38
rs1022352209 Chr.6: 106512563-106512563 on Build GRCh38
rs772497410 Chr.6: 106512593-106512593 on Build GRCh38
rs772051884 Chr.6: 106512410-106512410 on Build GRCh38
rs529525411 Chr.6: 106512159-106512159 on Build GRCh38
rs1031540328 Chr.6: 106512548-106512548 on Build GRCh38
rs771126437 Chr.6: 106512514-106512514 on Build GRCh38
rs762025410 Chr.6: 106512291-106512291 on Build GRCh38
rs775403138 Chr.6: 106512249-106512249 on Build GRCh38
rs776793620 Chr.6: 106512262-106512262 on Build GRCh38
rs766169689 Chr.6: 106512562-106512562 on Build GRCh38
rs1034781539 Chr.6: 106512293-106512293 on Build GRCh38
rs763362118 Chr.6: 106512309-106512309 on Build GRCh38
rs984324156 Chr.6: 106512365-106512365 on Build GRCh38
rs550678787 Chr.6: 106512317-106512317 on Build GRCh38
rs769649792 Chr.6: 106512210-106512210 on Build GRCh38
rs567384050 Chr.6: 106512342-106512342 on Build GRCh38
rs535274999 Chr.6: 106512361-106512361 on Build GRCh38
rs763541838 Chr.6: 106512468-106512468 on Build GRCh38
rs765458247 Chr.6: 106512296-106512296 on Build GRCh38
rs201789082 Chr.6: 106512507-106512507 on Build GRCh38
rs756154823 Chr.6: 106512501-106512501 on Build GRCh38
rs145352330 Chr.6: 106512407-106512407 on Build GRCh38
rs898158524 Chr.6: 106512208-106512208 on Build GRCh38
rs747536478 Chr.6: 106512595-106512595 on Build GRCh38
rs1022743186 Chr.6: 106512367-106512367 on Build GRCh38
rs146137130 Chr.6: 106512557-106512557 on Build GRCh38
rs753462793 Chr.6: 106512475-106512475 on Build GRCh38
rs768180047 Chr.6: 106512450-106512450 on Build GRCh38
rs777532627 Chr.6: 106512393-106512393 on Build GRCh38
rs776171328 Chr.6: 106512613-106512613 on Build GRCh38
rs201345513 Chr.6: 106512588-106512588 on Build GRCh38
rs918863563 Chr.6: 106512323-106512323 on Build GRCh38
rs763600737 Chr.6: 106512543-106512543 on Build GRCh38
rs749150119 Chr.6: 106512338-106512338 on Build GRCh38
rs149264191 Chr.6: 106512370-106512370 on Build GRCh38
rs748629269 Chr.6: 106512472-106512473 on Build GRCh38
rs748264514 Chr.6: 106512415-106512415 on Build GRCh38
rs537020222 Chr.6: 106512303-106512303 on Build GRCh38
rs746126802 Chr.6: 106512589-106512589 on Build GRCh38
rs749168690 Chr.6: 106512227-106512227 on Build GRCh38
rs763420956 Chr.6: 106512373-106512373 on Build GRCh38
rs752049592 Chr.6: 106512378-106512378 on Build GRCh38
rs750194381 Chr.6: 106512559-106512559 on Build GRCh38
rs200137698 Chr.6: 106512506-106512506 on Build GRCh38
rs114293299 Chr.6: 106512178-106512178 on Build GRCh38
rs778860060 Chr.6: 106512344-106512344 on Build GRCh38
rs767122979 Chr.6: 106512386-106512386 on Build GRCh38
rs769882846 Chr.6: 106512422-106512422 on Build GRCh38
rs758953735 Chr.6: 106512162-106512162 on Build GRCh38
rs763092727 Chr.6: 106512428-106512428 on Build GRCh38
rs777297975 Chr.6: 106512606-106512606 on Build GRCh38
rs934164410 Chr.6: 106512408-106512408 on Build GRCh38
rs758297422 Chr.6: 106512497-106512497 on Build GRCh38
rs756297940 Chr.6: 106512244-106512245 on Build GRCh38
rs1028540841 Chr.6: 106512599-106512599 on Build GRCh38
rs184750193 Chr.6: 106512250-106512250 on Build GRCh38
rs562698502 Chr.6: 106512447-106512447 on Build GRCh38
rs770969864 Chr.6: 106512235-106512235 on Build GRCh38
rs754500296 Chr.6: 106512327-106512327 on Build GRCh38

View Full Product Details