Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106520245-106520755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM1
Entrez Gene ID
202
Gene Name
absent in melanoma 1
Gene Aliases
CRYBG1, ST4
UniGene
Hs.724262
Gene Chromosome Location
Chr.6: 106360808-106570449 on Build GRCh38
Transcripts
NM_001624.3 XM_005266839.3 XM_017010333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857251 c.1869G>A p.P623P Chr.6: 106520301-106520301 on Build GRCh38
3619221 c.2092G>A p.E698K Chr.6: 106520524-106520524 on Build GRCh38
390288 c.2278C>T p.Q760* Chr.6: 106520710-106520710 on Build GRCh38
3154443 c.2020G>A p.D674N Chr.6: 106520452-106520452 on Build GRCh38
3154449 c.2312A>G p.Q771R Chr.6: 106520744-106520744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752787912 Chr.6: 106520580-106520580 on Build GRCh38
rs750404132 Chr.6: 106520300-106520300 on Build GRCh38
rs139190201 Chr.6: 106520585-106520585 on Build GRCh38
rs374531349 Chr.6: 106520559-106520559 on Build GRCh38
rs746629299 Chr.6: 106520377-106520377 on Build GRCh38
rs765446944 Chr.6: 106520641-106520641 on Build GRCh38
rs756548400 Chr.6: 106520445-106520445 on Build GRCh38
rs73761859 Chr.6: 106520494-106520494 on Build GRCh38
rs768856140 Chr.6: 106520635-106520635 on Build GRCh38
rs779223888 Chr.6: 106520361-106520361 on Build GRCh38
rs755546436 Chr.6: 106520687-106520687 on Build GRCh38
rs752201884 Chr.6: 106520702-106520702 on Build GRCh38
rs1004725274 Chr.6: 106520510-106520510 on Build GRCh38
rs771990517 Chr.6: 106520416-106520416 on Build GRCh38
rs761741079 Chr.6: 106520340-106520341 on Build GRCh38
rs778261644 Chr.6: 106520335-106520335 on Build GRCh38
rs756444706 Chr.6: 106520331-106520331 on Build GRCh38
rs780650499 Chr.6: 106520700-106520700 on Build GRCh38
rs375032575 Chr.6: 106520391-106520391 on Build GRCh38
rs763868734 Chr.6: 106520274-106520274 on Build GRCh38
rs749071520 Chr.6: 106520604-106520604 on Build GRCh38
rs750638973 Chr.6: 106520657-106520657 on Build GRCh38
rs779396471 Chr.6: 106520435-106520435 on Build GRCh38
rs41302206 Chr.6: 106520375-106520375 on Build GRCh38
rs371008588 Chr.6: 106520413-106520413 on Build GRCh38
rs763308337 Chr.6: 106520658-106520658 on Build GRCh38
rs754425255 Chr.6: 106520573-106520573 on Build GRCh38
rs761714288 Chr.6: 106520291-106520291 on Build GRCh38
rs143246482 Chr.6: 106520725-106520725 on Build GRCh38
rs912788634 Chr.6: 106520389-106520389 on Build GRCh38
rs370371694 Chr.6: 106520332-106520332 on Build GRCh38
rs993472966 Chr.6: 106520570-106520570 on Build GRCh38
rs757509100 Chr.6: 106520419-106520419 on Build GRCh38
rs144133205 Chr.6: 106520359-106520359 on Build GRCh38
rs748851621 Chr.6: 106520298-106520299 on Build GRCh38
rs755609344 Chr.6: 106520719-106520719 on Build GRCh38
rs779513280 Chr.6: 106520457-106520457 on Build GRCh38
rs775315490 Chr.6: 106520729-106520729 on Build GRCh38
rs765033889 Chr.6: 106520295-106520295 on Build GRCh38
rs374112542 Chr.6: 106520267-106520267 on Build GRCh38
rs1050137826 Chr.6: 106520709-106520709 on Build GRCh38
rs190627674 Chr.6: 106520477-106520477 on Build GRCh38
rs554277092 Chr.6: 106520491-106520491 on Build GRCh38
rs925671206 Chr.6: 106520626-106520626 on Build GRCh38
rs753669887 Chr.6: 106520285-106520285 on Build GRCh38
rs144739119 Chr.6: 106520411-106520411 on Build GRCh38
rs73520807 Chr.6: 106520330-106520330 on Build GRCh38
rs777415653 Chr.6: 106520721-106520721 on Build GRCh38
rs751433495 Chr.6: 106520590-106520590 on Build GRCh38
rs1016469700 Chr.6: 106520552-106520552 on Build GRCh38
rs967494729 Chr.6: 106520608-106520608 on Build GRCh38
rs373880356 Chr.6: 106520661-106520661 on Build GRCh38
rs530698683 Chr.6: 106520272-106520272 on Build GRCh38
rs1046336450 Chr.6: 106520499-106520499 on Build GRCh38
rs150468985 Chr.6: 106520301-106520301 on Build GRCh38
rs140546725 Chr.6: 106520273-106520273 on Build GRCh38
rs769462607 Chr.6: 106520485-106520485 on Build GRCh38
rs200098719 Chr.6: 106520727-106520727 on Build GRCh38
rs772501819 Chr.6: 106520599-106520599 on Build GRCh38
rs771473917 Chr.6: 106520374-106520374 on Build GRCh38
rs925638609 Chr.6: 106520390-106520390 on Build GRCh38
rs768183698 Chr.6: 106520461-106520461 on Build GRCh38
rs760490539 Chr.6: 106520584-106520586 on Build GRCh38
rs776198080 Chr.6: 106520463-106520463 on Build GRCh38
rs776284327 Chr.6: 106520379-106520379 on Build GRCh38
rs745764944 Chr.6: 106520726-106520726 on Build GRCh38
rs147638688 Chr.6: 106520318-106520319 on Build GRCh38
rs112245852 Chr.6: 106520507-106520507 on Build GRCh38
rs746469912 Chr.6: 106520458-106520458 on Build GRCh38
rs758103468 Chr.6: 106520607-106520607 on Build GRCh38
rs767465093 Chr.6: 106520397-106520397 on Build GRCh38
rs866093812 Chr.6: 106520327-106520327 on Build GRCh38
rs770833441 Chr.6: 106520724-106520724 on Build GRCh38
rs781344637 Chr.6: 106520325-106520325 on Build GRCh38
rs747848536 Chr.6: 106520483-106520483 on Build GRCh38
rs752063356 Chr.6: 106520572-106520572 on Build GRCh38
rs141256151 Chr.6: 106520558-106520558 on Build GRCh38
rs113287212 Chr.6: 106520569-106520569 on Build GRCh38
rs201201445 Chr.6: 106520398-106520398 on Build GRCh38
rs150778380 Chr.6: 106520560-106520560 on Build GRCh38
rs143063193 Chr.6: 106520386-106520386 on Build GRCh38
rs535796320 Chr.6: 106520439-106520440 on Build GRCh38
rs139061896 Chr.6: 106520381-106520381 on Build GRCh38
rs894785850 Chr.6: 106520467-106520467 on Build GRCh38
rs199579020 Chr.6: 106520611-106520611 on Build GRCh38
rs748904708 Chr.6: 106520723-106520723 on Build GRCh38
rs774375182 Chr.6: 106520615-106520615 on Build GRCh38
rs749622666 Chr.6: 106520339-106520339 on Build GRCh38
rs774462967 Chr.6: 106520500-106520500 on Build GRCh38
rs747751309 Chr.6: 106520586-106520586 on Build GRCh38
rs761914015 Chr.6: 106520640-106520640 on Build GRCh38
rs758980135 Chr.6: 106520454-106520454 on Build GRCh38
rs768173586 Chr.6: 106520300-106520301 on Build GRCh38
rs773126523 Chr.6: 106520490-106520490 on Build GRCh38
rs758931666 Chr.6: 106520271-106520271 on Build GRCh38
rs754101895 Chr.6: 106520553-106520553 on Build GRCh38
rs867712379 Chr.6: 106520438-106520438 on Build GRCh38
rs746403735 Chr.6: 106520362-106520362 on Build GRCh38
rs764284968 Chr.6: 106520539-106520539 on Build GRCh38
rs768343143 Chr.6: 106520378-106520378 on Build GRCh38
rs370780499 Chr.6: 106520619-106520619 on Build GRCh38
rs776895259 Chr.6: 106520636-106520636 on Build GRCh38
rs367560294 Chr.6: 106520589-106520589 on Build GRCh38
rs777313852 Chr.6: 106520596-106520596 on Build GRCh38
rs768045613 Chr.6: 106520516-106520516 on Build GRCh38
rs752049159 Chr.6: 106520674-106520674 on Build GRCh38

View Full Product Details